CLS7095D9
Corning® disposable Pasteur pipettes, bulk pack, non-sterile, unplugged
size 9 in., borosilicate
Synonym(s):
Pipettes, pipettes, serological pipets, serological pipettes
All Photos(1)
About This Item
UNSPSC Code:
41121500
eCl@ss:
32079210
NACRES:
NB.04
Recommended Products
material
borosilicate glass
sterility
non-sterile
packaging
pack of 200 ea
manufacturer/tradename
Corning 7095D-9
size
9 in.
General description
- Corning® disposable Pasteur pipettes are made of sturdy and optically clear borosilicate glass.
- Single-use pipettes prevent the risk of cross-contamination of valuable specimens or laboratory reagents.
- Feature a constriction one inch below the top to facilitate plugging.
- Ideal for use in non-volumetric transfer work in bacteriology, immunology, hematology, and serology studies, as well as blood bank and clinical chemistry procedures.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)