CLS4143
Corning® DeckWorks™ pipet tip station
0.1 - 10μL pipet tip station, extended length, graduated, sterile, polypropylene, 192 tips/layer, 5 layers/pack, 5 packs/case
Synonym(s):
pipet tip station, pipette tip station
About This Item
RNase free
barrier: no
disposable
endotoxin free
filter: no
graduated
Tip shape: round
Recommended Products
material
natural tip
polypropylene
sterility
sterile
feature
DNase free
RNase free
barrier: no
disposable
endotoxin free
filter: no
graduated
Tip shape: round
packaging
pack of 960
manufacturer/tradename
Corning 4143
volume range
0.1-10 μL
volume
0.1—10 μL
suitability
suitable for (pre- and post-PCR procedures, cell culture and assays requiring large tip quantities)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Corning DeckWorks tip station maximizes bench space while minimizing plastic waste. Tip stations contain 960 tips, are available in extended length 10 uL and traditional 200 uL sizes, and are the ideal single and multi-channel pipetting stations. The 5-layer deck design (2 x 96) matches the microtiter plate format. This stable pipetting platform fits easily into sterile hood environments. Lids can be snap-positioned to expose a single side (96 tips) only.
Applications include pre- and post-PCR procedures, cell culture and assays requiring large tip quantities.
Corning DeckWorks tip stations are offered both sterile and nonsterile. This system is designed to allow you to maximize storage space and help reduce the production and waste of plastic materials. Both the hinged racks and reload decks within this system are manufactured with recycled plastic.
The pipet tips are made in a controlled molding environment to ensure accuracy and precision across all tip volumes.
Medical grade resins and production excellence allow for full certification as:
- Nonpyrogenic
- Human DNA-free
- RNase-/DNase-free
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service