Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
DeckWorks
™
pipet tip station
Products
Product Category
pipette tips (4)
Manufacturer
Volume Range (µL)
Color
Package Size
Sterility
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
CLS4144
Corning
®
DeckWorks
™
pipet tip station,
1 - 200μL pipet tip station, graduated, sterile, polypropylene, 192 tips/layer, 5 layers/pack, 5 packs/case
Expand
View Pricing
CLS4142
Corning
®
DeckWorks
™
pipet tip station,
1 - 200μL pipet tip station, graduated, non-sterile, polypropylene, 192 tips/layer, 5 layers/pack, 5 packs/case
Expand
View Pricing
CLS4143
Corning
®
DeckWorks
™
pipet tip station,
0.1 - 10μL pipet tip station, extended length, graduated, sterile, polypropylene, 192 tips/layer, 5 layers/pack, 5 packs/case
Expand
View Pricing
CLS4141
Corning
®
DeckWorks
™
pipet tip station,
0.1 - 10μL pipet tip station, extended length, graduated, non-sterile, polypropylene, 192 tips/layer, 5 layers/pack, 5 packs/case
Expand
View Pricing