Skip to Content
Merck
All Photos(1)

Key Documents

CLS3764

Corning® 384 well microplate, low flange

Tissue Culture (TC)-treated surface, black polystyrene, flat bottom, sterile, lid, pack of 20

Synonym(s):

384 multiwell plates, 384 well immunoassay plates, 384 well microplates, 384 well microtiter plates, 384 well plates, immunoassay plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.24
material:
black polystyrene
clear bottom
flat bottom
flat wells
polystyrene
polystyrene plate
maximum volume:
112 μL
size:
384 wells
384 wells
well volume:
112 μL
well working volume:
25-50 μL
working volume:
80 μL

material

black polystyrene
clear bottom
flat bottom
flat wells
polystyrene
polystyrene plate

sterility

sterile

feature

lid
skirt
plate format: low flange

packaging

case of 100 ea
pack of 20

manufacturer/tradename

Corning 3764

maximum volume

112 μL

size

384 wells
384 wells

well volume

112 μL

well working volume

25-50 μL

working volume

80 μL

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Corning® 384 well microplate, low flange features a clear and flat well bottom with a black wall. It is designed with an enhanced molding process that reduces autofluorescence and unwanted background signals leading to lower coefficient of variations (CVs), ensuring reliable and accurate assay results. Additionally, the plate has barcode customization and robotic handling. It is well-suited for conducting highly sensitive assays and is used in microplate readers efficiently.

Application

Corning® 384 well microplate, low flange has been used in resazurin assay and high throughput RNA interference screening.

Features and Benefits

  • Clear flat well bottom with a black wall
  • Non-pyrogenic, sterilized by gamma radiation
  • Tissue culture-treated plates for better cell attachment
  • have opaque walls that exhibit low background fluorescence, reducing light scattering and minimizing crosstalk between wells.
  • Allows direct microscopic viewing due to the presence of flat bottom with an optically clear well
  • Used as reading instruments for both top and bottom reading
  • Features individual alphanumeric codes, allowing for easy well identification.
  • Provides 20% higher signal-to-background ratio resulting in enhanced assay sensitivity.
  • The plates offer superior bottom flatness (~100 µm) that reduces well-to-well variation and provides larger growth area for cell-based assays.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MTH1 as a target to alleviate T cell driven diseases by selective suppression of activated T cells
Stella Karsten
Cell Death and Differentiation, 29, 246-261 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service