Skip to Content
Merck
All Photos(1)

Key Documents

CLS3680

Corning® 384 well plates

lid, sterile; γ-irradiated

Synonym(s):

384 multiwell plates, 384 well low volume plates, 384 well microplates, 384 well microtiter plates, 384 well plates, 384 well tissue culture treated plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.24
material:
clear polystyrene plate
flat bottom clear wells
polystyrene
square clear wells
maximum volume:
112 μL
size:
384 wells
384 wells
working volume:
80 μL

material

clear polystyrene plate
flat bottom clear wells
polystyrene
square clear wells

sterility

sterile; γ-irradiated

feature

lid
skirt

packaging

case of 100

manufacturer/tradename

Corning 3680

maximum volume

112 μL

size

384 wells
384 wells

working volume

80 μL

color

clear

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

For optical imaging
These 384 well plates are compatible with leading readers and integrate into high-throughput applications.

Features and Benefits

  • Corning® 384 well plates are ELISA microplates with a working volume of up to 20 to 80 μL
  • Standard microplate footprint and dimensions
  • Not treated polystyrene surface
  • Hydrophobic surface binds biomolecules by passive interactions
  • Applicable for the immobilization of large molecules
  • Untreated microplates have a binding capacity of approximately 100 to 200ng IgG/cm²

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angelo Frei et al.
Chemical science, 11(10), 2627-2639 (2020-03-25)
There is a dire need for new antimicrobial compounds to combat the growing threat of widespread antibiotic resistance. With a currently very scarce drug pipeline, consisting mostly of derivatives of known antibiotics, new classes of antibiotics are urgently required. Metal
Angelo Frei et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 27(6), 2021-2029 (2020-11-25)
Resistance to currently available antifungal drugs has quietly been on the rise but overshadowed by the alarming spread of antibacterial resistance. There is a striking lack of attention to the threat of drug-resistant fungal infections, with only a handful of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service