Skip to Content
Merck
All Photos(1)

Key Documents

CLS3363

Corning® 96 Well Storage Microplate

V-bottom clear, polypropylene, bag of 25 ×, non-sterile, lid: no

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
V-bottom clear
polypropylene
maximum volume:
320 μL
size:
96 wells
working volume:
200 μL
Pricing and availability is not currently available.

material

V-bottom clear
polypropylene

sterility

non-sterile

feature

lid: no

packaging

case of 100
bag of 25 ×

manufacturer/tradename

Corning 3363

maximum volume

320 μL

size

96 wells

working volume

200 μL

color

clear

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Feature uniform skirt heights for greater robotic gripping surface
  • Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
  • Certified DNase- and RNase-free
  • Total volume is 320 μL; recommended working well volume is 75 to 200 μL
  • Without lids; lids, sealing mats and tapes supplied separately

Features and Benefits

  • Corning® 96 well microplates support various assay and storage application requirements.
  • Features uniform skirt heights for the greater robotic gripping surface.
  • Resistant to most solvents including DMSO, ethanol, methanol.
  • RNase-/DNase-free.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service