Skip to Content
Merck
All Photos(1)

Key Documents

CLS3080

Corning ® 96 Well Microplate Storage Mat

non-sterile

Synonym(s):

96-Well Storage Mat, Microplate Storage Mat, Storage Mat for Microplates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
size:
96 wells
Pricing and availability is not currently available.

description

storage mat III

sterility

non-sterile
non-sterile

packaging

case of 100 ea
pack of 25 ea

manufacturer/tradename

Corning 3080

size

96 wells

suitability

suitable for (general storage, liquid handling; automated applications)

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Corning® 96 Well Microplate Storage Mat fits Corning® 96 round well plates and blocks.
  • Manufactured from EVA (ethyl vinyl acetate) and is compatible with DMSO.
  • Nonsterile and non-treated surface.
  • Free of DNase- and RNase.
  • Applied manually or with Storage Mat Applicator.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service