Key Documents
CLS2980246
Corning® cover glasses
rectangular, No. 11/2, W × L 24 mm × 60 mm
Synonym(s):
coverslips, glass covers, slide covers
About This Item
UNSPSC Code:
41121800
NACRES:
NB.16
Pricing and availability is not currently available.
material
borosilicate glass
rectangular
packaging
pack of 100 ea
case of 1000 ea
manufacturer/tradename
Corning 2980-246
W × L
24 mm × 60 mm
binding type
non-treated surface
Features and Benefits
  • Corning® cover glasses are made from optically clear borosilicate glass.
  • Chemically resistant cover glasses are not affected by weathering or surface attachment.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)