Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
cover glasses
Products
Manufacturer
Binding Type
Package Size
Filter Products
Showing 1-20 of 23
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
CLS2980245
Corning
®
cover glasses,
rectangular, No. 1
1
/
2
, W × L 24 mm × 50 mm
Expand
View Pricing
CLS284518
Corning
®
cover glasses,
square, No. 1, W x L 18 mm x 18 mm
Expand
View Pricing
CLS2975245
Corning
®
cover glasses,
rectangular, No. 1, W × L 24 mm × 50 mm
Expand
View Pricing
CLS2975246
Corning
®
cover glasses,
rectangular, No. 1, W × L 24 mm × 60 mm
Expand
View Pricing
CLS2975225
Corning
®
cover glasses,
rectangular, No. 1, W × L 22 mm × 50 mm
Expand
View Pricing
CLS284522
Corning
®
cover glasses,
square, No. 1, W x L 22 mm x mm
Expand
View Pricing
CLS2980246
Corning
®
cover glasses,
rectangular, No. 1
1
/
2
, W × L 24 mm × 60 mm
Expand
View Pricing
CLS2980225
Corning
®
cover glasses,
rectangular, No. 1
1
/
2
, W × L 22 mm × 50 mm
Expand
View Pricing
CLS285525
Corning
®
cover glasses,
square, No. 2, W × L 25 mm × 25 mm
Expand
View Pricing
CLS2975244
Corning
®
cover glasses,
rectangular, No. 1, W × L 24 mm × 40 mm
Expand
View Pricing
CLS285518
Corning
®
cover glasses,
square, No. 2, W × L 18 mm × 18 mm
Expand
View Pricing
CLS2975224
Corning
®
cover glasses,
rectangular, No. 1, W × L 22 mm × 40 mm
Expand
View Pricing
CLS2975223
Corning
®
cover glasses,
rectangular, No. 1, W × L 22 mm × 30 mm
Expand
View Pricing
CLS285022
Corning
®
cover glasses,
square, No. 1
1
/
2
, W × L 22 mm × 22 mm
Expand
View Pricing
CLS285018
Corning
®
cover glasses,
square, No. 1
1
/
2
, W × L 18 mm × 18 mm
Expand
View Pricing
CLS2980224
Corning
®
cover glasses,
rectangular, No. 1
1
/
2
, W × L 22 mm × 40 mm
Expand
View Pricing
CLS284525
Corning
®
cover glasses,
square, No. 1, W x L 25 mm x 25 mm
Expand
View Pricing
CLS285522
Corning
®
cover glasses,
square, No. 2, W × L 22 mm × 22 mm
Expand
View Pricing
CLS285025
Corning
®
cover glasses,
square, No. 1
1
/
2
, W × L 25 mm × 25 mm
Expand
View Pricing
CLS2980223
Corning
®
cover glasses,
rectangular, No. 1
1
/
2
, W × L 22 mm × 30 mm
Expand
View Pricing
Page 1 of 2
Page 1 of 2