Recommended Products
description
CHOZN® GS-/- ZFN-modified CHO cell line
glutamine synthetase knockout cell line
solubility
DMSO: 5 mg/mL, clear
shipped in
liquid nitrogen
storage temp.
−196°C
Looking for similar products? Visit Product Comparison Guide
General description
CHOZN cell lines were created utilizing Sigma′s proprietary CompoZr Zinc Finger Nuclease (ZFN) technology to create genetic modifications in parental CHO-K1 cells. Cells are banked in chemically defined, animal component free EX-CELL CD CHO Fusion media
ZFNs are a class of engineered DNA-binding proteins and causes a double-strand break. This double-strand break is necessary for site-specific mutagenesis mediated DNA repair. It facilitates targeted genome editing by binding to a user-specified locus. Glutamine synthetase (GS) is one of the most commonly used selectable markers in the biopharmaceutical industry. Glutamine is an essential amino acid for cellular growth. The GS enzyme is responsible for the conversion of glutamate into glutamine. Without this enzymatic activity, cells can no longer synthesize glutamine endogenously and if glutamine is not supplemented in the culture media, the cells will die.
Cell Line Origin
CHOZN cell lines are all subclones of the parental CHO-K1 cell line originated by Puck in 1957.
Packaging
This product is shipped in liquid nitrogen dry vapor shippers.
Liquid nitrogen dry vapor shipper fee (up to a maximum of $125) and freight not included in price of product.
Liquid nitrogen dry vapor shipper is not disposable and should be returned after receipt. A $450 liquid nitrogen dry vapor shipper fee will be charged if not returned within 1 week of delivery.
Learn more about liquid nitrogen dry vapor shipper
Liquid nitrogen dry vapor shipper fee (up to a maximum of $125) and freight not included in price of product.
Liquid nitrogen dry vapor shipper is not disposable and should be returned after receipt. A $450 liquid nitrogen dry vapor shipper fee will be charged if not returned within 1 week of delivery.
Learn more about liquid nitrogen dry vapor shipper
Physical form
CHOZN cell lines are provided to customers in vials containing >7.5e6 cells/mL
Legal Information
All CHOZN cell lines are sold under license from Sigma Aldrich. A copy of the research license is provided with purchase of the CHOZN cell lines.
CHOZN is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Q's next: the diverse functions of glutamine in metabolism, cell biology and cancer.
DeBerardinis R J and Cheng T
Oncogene, 29(3), 313-313 (2010)
Cytotoxic effects during knock out of multiple porcine endogenous retrovirus (PERV) sequences in the pig genome by zinc finger nucleases (ZFN).
Semaan M, et al.
PLoS ONE, 10(4), e0122059-e0122059 (2015)
The metabolic profile of tumors depends on both the responsible genetic lesion and tissue type.
Yuneva M O, et al.
Cell Metabolism, 15(2), 157-170 (2012)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service