Skip to Content
Merck
All Photos(2)

Key Documents

C8552

Sigma-Aldrich

Cytidine 5′-triphosphate disodium solution

HPLC purified, aqueous solution for RNA polymerase transcription

Synonym(s):

CTP-Na2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16N3O14P3Na2
CAS Number:
Molecular Weight:
529.14
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.52
Pricing and availability is not currently available.

Quality Level

form

liquid

concentration

95-105 mM in H2O (adjusted to pH 7 with Sodium Hydroxide)

color

colorless

foreign activity

DNase and RNase, none detected

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]2O

InChI

1S/C9H16N3O14P3.2Na/c10-5-1-2-12(9(15)11-5)8-7(14)6(13)4(24-8)3-23-28(19,20)26-29(21,22)25-27(16,17)18;;/h1-2,4,6-8,13-14H,3H2,(H,19,20)(H,21,22)(H2,10,11,15)(H2,16,17,18);;/q;2*+1/p-2/t4-,6-,7-,8-;;/m1../s1

InChI key

NFQMDTRPCFJJND-WFIJOQBCSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Cytidine 5′-triphosphate (CTP) is a high-energy molecule and pyrimidine nucleoside triphosphate. It acts as a substrate for the RNA synthesis. CTP also acts as a coenzyme during the synthesis of glycerophospholipids and glycosylation of proteins. It is made up of cytidine and three phosphate groups and can be a potent phosphate source.[1]

Application

For use in DNA-dependent RNA polymerase transcription

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sambrook, J., et al.
Molecular Cloning: A Laboratory Manual, 5-5 (1989)
Microwave-assisted hydrothermal rapid synthesis of amorphous calcium phosphate nanoparticles and hydroxyapatite microspheres using cytidine 5'-triphosphate disodium salt as a phosphate source
Zhao J, et al.
Materials Letters, 124, 208-211 (2014)
Xi Liu et al.
Anti-cancer drugs, 25(6), 690-703 (2014-03-29)
Treatment of mammalian cells with chemotherapeutic drugs can result in perturbations of nucleotide pools. Monitoring these perturbations in cultured tumor cells from human sources is useful for assessment of the effect of drug therapy and a better understanding of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service