Product C7231 has a filter screw cap
C7231
Greiner culture flasks, tissue culture treated
surface area 75 cm2, canted neck, with cap, filtered
Synonym(s):
culture flask, tissue culture flask, tissue culture flasks
About This Item
Recommended Products
material
canted neck
polystyrene flask
screw top red high-density polyethylene cap
sterility
sterile
feature
DNase free
RNase free
with cap filtered
packaging
pack of 120 ea
manufacturer/tradename
Greiner 658175
capacity
250 mL
surface area
75 cm2
working volume
38 mL
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Greiner culture flasks offer optimal cell adhesion through physical surface treatment.
- Flasks are non-cytotoxic, non-pyrogenic and free of detectable DNase, RNase and human DNA.
- The specific design facilitates optimal access to the samples with a cell scraper or pipette.
- The canted neck ensures additional protection against contamination.
- A standard screw cap with a secured click-in ventilation position enables gas exchange within the flasks.
NOTE: For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Does Product C7231 come as plug sealed or vented?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service