Skip to Content
Merck
All Photos(3)

Key Documents

F4381

Sigma-Aldrich

Furosemide

≥98% (HPLC), powder, NKCC symporter inhibitor

Synonym(s):

4-Chloro-N-furfuryl-5-sulfamoylanthranilic acid, 5-(Aminosulfonyl)-4-chloro-2-([2-furanylmethyl]amino)benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11ClN2O5S
CAS Number:
Molecular Weight:
330.74
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Furosemide,

biological source

synthetic (organic)

Assay

≥98% (HPLC)

form

powder

mp

220 °C (dec.) (lit.)

solubility

acetone: 50 mg/mL, clear to slightly hazy, colorless to yellow

originator

Sanofi Aventis

storage temp.

room temp

SMILES string

NS(=O)(=O)c1cc(C(O)=O)c(NCc2ccco2)cc1Cl

InChI

1S/C12H11ClN2O5S/c13-9-5-10(15-6-7-2-1-3-20-7)8(12(16)17)4-11(9)21(14,18)19/h1-5,15H,6H2,(H,16,17)(H2,14,18,19)

InChI key

ZZUFCTLCJUWOSV-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Furosemide is a an ototoxic high-ceiling diuretic.

Application

Furosemide has been used:
  • to investigate the influence of timing of dexamethasone administration on auditory hair cell survival in mice
  • to study its effect on intracranial pressure (ICP) after subcutaneous administration in rats
  • as a stimulant for renin release and subsequent increase in plasma angiotensin II (ANG II)

Biochem/physiol Actions

"High ceiling" diuretic that strongly affects renal tubular action by increasing renal blood flow; antihypertensive.
Furosemide can block the Na+/K+/2Cl- co-transporter in the ascending thick loop of Henle. It can enhance the synthesis of intrarenal prostaglandin. It enhances its ototoxicity in animals when used along with kanamycin. Furosemide is linked with thiamine insufficiency in individuals with heart failure.
Inhibits ion co-transport in the kidney.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Sanofi Aventis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diuretics
Side Effects of Drugs Annual, 28, 233-243 (2005)
Topiramate is more effective than acetazolamide at lowering intracranial pressure
Scotton W J, et al.
Cephalalgia, 0333102418776455-0333102418776455 (2018)
Furosemide
Saunders Handbook of Veterinary Drugs - E-Book (2016)
Causes of acquired hearing loss
Eggermont J J
Hearing Loss: Causes, Prevention, and Treatment (2017)
Development of an ototoxicity model in the adult CBA/CaJ mouse and determination of a golden window of corticosteroid intervention for otoprotection
Fernandes V T and Lin V YW
Journal of Otolaryngology - Head & Neck Surgery = Le Journal D'Oto-rhino-laryngologie et de Chirurgie Cervico-Faciale, 43(1), 12-12 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service