Skip to Content
Merck
All Photos(1)

Documents

C4356

Sigma-Aldrich

Anti-Caspase 9 antibody,Mouse monoclonal

clone CAS9, purified from hybridoma cell culture

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

CAS9, monoclonal

form

buffered aqueous solution

mol wt

antigen 37 kDa

species reactivity

human

concentration

~2 mg/mL

technique(s)

indirect ELISA: suitable
microarray: suitable
western blot: 0.5-1 μg/mL using a purified recombinant human caspase 9 preparation

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CASP9(842)

General description

Monoclonal Anti-Caspase 9 (mouse IgG1 isotype) is derived from the CAS9 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from a BALB/c mouse immunized with a synthetic peptide corresponding to amino acids of human caspase 9 protein, conjugated to keyhole limpet hemocyanin (KLH). Caspases (cysteine-requiring aspartate protease) are a family of proteases that share similarities in amino acid sequences, structure and substrate specificity. Caspases are normally present in the cell as inactive procaspases. Caspase 9 (also known as Mch6 and ICE-LAP6) gene encodes a protein product of 37 kDa (reported also as 46 kDa), which undergoes a cleavage upon activation, leading to p18/p15/p9 subunits. It is a member of the CED-3 subfamily, bearing high similarity to caspase. High level expression of caspase 9 is found in the heart, testis and ovary.

Immunogen

synthetic peptide corresponding to amino acids 299-316 of human caspase 9 protein conjugated to KLH.

Application

Anti-Caspase 9 antibody,Mouse monoclonal has been used in:
  • western blot assay
  • immunocytochemistry
  • enzyme-linked immunosorbent assay (ELISA).

Biochem/physiol Actions

Caspase 9 is a downstream protease, activated in caspase 3- and granzyme B-mediated apoptosis. Procaspase 9 is activated when a complex is formed with the cofactor apoptotic protease activating factor-1 (Apaf-1) through the caspase recruitment domain (CARD). Caspase 9 plays a critical role in the survival of neuronal progenitors within the embryonic forebrain and cortex. Its deficiency leads to defective brain development and embryonic lethality. Overexpression of caspase 9, but not of a mutant in which the catalytic Cys was replaced with an Ala, induces apoptosis in MCF7 cells.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

CD 47 blockade reduces ischemia/reperfusion injury in donation after cardiac death rat kidney transplantation
Wang X, et al.
American Journal of Transplantation : Official Journal of the American Society of Transplantation and the American Society of Transplant Surgeons, 18(4), 843-854 (2018)
Role of caspases in the regulation of apoptotic pancreatic islet beta-cells death
Hui H, et al.
Journal of Cellular Physiology, 200(2), 177-200 (2004)
Apoptosis dependent and independent functions of caspases
Madame Curie Bioscience Database [Internet]. (2013)
Generation of Advanced Glycation End-Products (AGEs) by glycoxidation mediated by copper and ROS in a human serum albumin (HSA) model peptide: reaction mechanism and damage in motor neuron cells
Marques CMS, et al.
Mutation Research. Genetic Toxicology and Environmental Mutagenesis, 824(6), 42-51 (2017)
Neural-specific inactivation of ShcA results in increased embryonic neural progenitor apoptosis and microencephaly
McFarland KN, et al.
The Journal of Neuroscience, 26(30), 7885-7897 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service