Skip to Content
Merck
All Photos(4)

Documents

C3915

Sigma-Aldrich

Colchicine

BioReagent, suitable for plant cell culture, ≥95% (HPLC)

Synonym(s):

(S)-N-(5,6,7,9-Tetrahydro-1,2,3,10-tetramethoxy-9-oxobenzo[a]heptalen-7-yl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H25NO6
CAS Number:
Molecular Weight:
399.44
Beilstein:
2228813
EC Number:
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72

product line

BioReagent

Assay

≥95% (HPLC)

form

powder

technique(s)

cell culture | plant: suitable

mp

150-160 °C (dec.) (lit.)

application(s)

agriculture

SMILES string

COC1=CC=C2C(=CC1=O)[C@H](CCc3cc(OC)c(OC)c(OC)c23)NC(C)=O

InChI

1S/C22H25NO6/c1-12(24)23-16-8-6-13-10-19(27-3)21(28-4)22(29-5)20(13)14-7-9-18(26-2)17(25)11-15(14)16/h7,9-11,16H,6,8H2,1-5H3,(H,23,24)/t16-/m0/s1

InChI key

IAKHMKGGTNLKSZ-INIZCTEOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Colchicine is a tricyclic alkaloid that can stay in circulation for 20 to 40 hours post intake. Leucocytes harbour maximum colchicine. It contains three hexameric rings termed A, B and C.

Application

Colchicine has been used:
  • as an inhibitor of tubulin polymerization in bone-marrow-derived macrophages
  • for the inhibition of in vitro neurite outgrowth in human neuronal cells
  • as microtubule destabilizer to induce peripheral neuropathy in induced pluripotent stem cell derived neurons (iPSC-Neurons)

Biochem/physiol Actions

Colchicine interacts with albumin and binds to tubulin. Its association with tubulin impacts autophagic vacuole fusion with lysosomes. It inhibits tyrosine kinases and phospholipases. Colchicine may be useful for treating acute coronary syndromes. It is prescribed for treating rheumatologic conditions including familial mediterranean fever (FMF) and acute gouty arthritis.
Colchicine is an inhibitor of microtubule polymerization which blocks chromosome segregation during meiosis. Consequently, Colchicine is used to induce polyploidy (tetraploid) in plant cells.

Preparation Note

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Muta. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effects of colchicine and hydroxychloroquine on the cyclo-oxygenases COX-1 and COX-2
Ben-Chetrit E, et al.
Rheumatology International, 25(5), 332-335 (2005)
NEK7 is an essential mediator of NLRP3 activation downstream of potassium efflux
He Y, et al.
Nature, 530(7590), 354-354 (2016)
Colchicine: old and new
Slobodnick A, et al.
The American Journal of Medicine, 128(5), 461-470 (2015)
Mechanism of the anti-inflammatory effect of colchicine in rheumatic diseases: a possible new outlook through microarray analysis
Ben-Chetrit E, et al.
Rheumatology (Basel), 45(3), 274-282 (2005)
Comparative Sensitivity of Human-Induced Pluripotent Stem Cell-Derived Neuronal Subtypes to Chemically Induced Neurodegeneration
Cohen JD and Tanaka Y
Applied In Vitro Toxicology, 4(4), 347-364 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service