Skip to Content
Merck

D1925

Demecolcine solution

10 μg/mL in HBSS, ACF Qualified, BioXtra

Synonym(s):

Colcemid solution, N-Deacetyl-N-methylcolchicine solution

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

CAS Number:
NACRES:
NA.75
PubChem Substance ID:
UNSPSC Code:
12352207
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Demecolcine solution, 10 μg/mL in HBSS, ACF Qualified, BioXtra

InChI key

NNJPGOLRFBJNIW-HNNXBMFYSA-N

SMILES string

CN[C@H]1CCc2cc(OC)c(OC)c(OC)c2C3=CC=C(OC)C(=O)C=C13

InChI

1S/C21H25NO5/c1-22-15-8-6-12-10-18(25-3)20(26-4)21(27-5)19(12)13-7-9-17(24-2)16(23)11-14(13)15/h7,9-11,15,22H,6,8H2,1-5H3/t15-/m0/s1

sterility

sterile-filtered

product line

BioXtra

form

liquid

concentration

10 μg/mL in HBSS

technique(s)

cell culture | mammalian: suitable

storage temp.

2-8°C

Quality Level

Looking for similar products? Visit Product Comparison Guide

Application

Used for cell synchronization by arresting them at metaphase. Qualified for use in animal component free applications.

Legal Information

Colcemid is a trademark of Ciba-Geigy Corporation

Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vandana Relan et al.
PloS one, 8(3), e58132-e58132 (2013-03-22)
Malignant mesothelioma is an aggressive tumour of serosal surfaces most commonly pleura. Characterised cell lines represent a valuable tool to study the biology of mesothelioma. The aim of this study was to develop and biologically characterise six malignant mesothelioma cell
Ting-Yuan Tseng et al.
Molecules (Basel, Switzerland), 25(18) (2020-09-11)
Identification of the existence of G-quadruplex (G4) structure, from a specific G-rich sequence in cells, is critical to the studies of structural biology and drug development. Accumulating evidence supports the existence of G4 structure in vivo. Particularly, time-gated fluorescence lifetime
Dimitris Ioannou et al.
Chromosome research : an international journal on the molecular, supramolecular and evolutionary aspects of chromosome biology, 17(4), 519-530 (2009-08-01)
In the field of nanotechnology, quantum dots (QDs) are a novel class of inorganic fluorochromes composed of nanometre-scale crystals made of a semiconductor material. Given the remarkable optical properties that they possess, they have been proposed as an ideal material
Peter Burkovics et al.
Nucleic acids research, 44(7), 3176-3189 (2016-01-23)
Successful and accurate completion of the replication of damage-containing DNA requires mainly recombination and RAD18-dependent DNA damage tolerance pathways. RAD18 governs at least two distinct mechanisms: translesion synthesis (TLS) and template switching (TS)-dependent pathways. Whereas TS is mainly error-free, TLS
Norio Hamada et al.
PLoS genetics, 16(3), e1008676-e1008676 (2020-03-28)
A set of sex chromosomes is required for gametogenesis in both males and females, as represented by sex chromosome disorders causing agametic phenotypes. Although studies using model animals have investigated the functional requirement of sex chromosomes, involvement of these chromosomes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service