Skip to Content
Merck
All Photos(1)

Key Documents

C2992

Sigma-Aldrich

CFTR Inhibitor-172

≥98% (HPLC), powder, CFTR inhibitor

Synonym(s):

5-[(4-Carboxyphenyl)methylene]-2-thioxo-3-[(3-trifluoromethyl)phenyl-4-thiazolidinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H10F3NO3S2
CAS Number:
Molecular Weight:
409.40
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

CFTR(inh)-172, ≥98% (HPLC), powder

Quality Level

Assay

≥98% (HPLC)

form

powder

color

yellow

solubility

DMSO: ≥10 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

OC(=O)c1ccc(cc1)\C=C2\SC(=S)N(C2=O)c3cccc(c3)C(F)(F)F

InChI

1S/C18H10F3NO3S2/c19-18(20,21)12-2-1-3-13(9-12)22-15(23)14(27-17(22)26)8-10-4-6-11(7-5-10)16(24)25/h1-9H,(H,24,25)/b14-8+

InChI key

JIMHYXZZCWVCMI-RIYZIHGNSA-N

Application

CFTR(inh)-172 has been used to specifically inhibit cystic fibrosis transmembrane conductance regulator (CFTR) activity.[1][2] It is also used as a CFTR inhibitor Inh-172 to treat forskolin and IBMX.[3]

Biochem/physiol Actions

CFTR(inh)-172 is an inhibitor of the cystic fibrosis transmembrane conductance regulator (CFTR).
CFTR(inh)-172 is an inhibitor of the cystic fibrosis transmembrane conductance regulator (CFTR). With a Ki = 300 nM,. CFTR(inh)-172 leads to rapid, reversible and voltage-independent inhibition; it is an antidiarrheal agent in animals. CTFR(inh)-172 may be a useful tool to study animal models of cystic fibrosis and intestinal fluid loss in cholera and other secretory diarrheas. CTFR(inh)-172 is structurally-unrelated to known, but non-specific CFTR inhibitors DPC, NPPB (Cat. No. N4779) and Glibenclamide.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N D Sonawane et al.
Journal of pharmaceutical sciences, 94(1), 134-143 (2005-03-12)
A small-molecule inhibitor of the cystic fibrosis transmembrane conductance regulator (CFTR), 3-[(3-trifluoromethyl)phenyl]-5-[(4-carboxyphenyl)methylene]-2-thioxo-4-thiazolidinone (CFTR(inh)-172), reduces enterotoxin-induced intestinal fluid secretion in rodents. Here, we study CFTR(inh)-172 pharmacology and antidiarrheal efficacy in rodents using (14)C-labeled CFTR(inh)-172, liquid chromatography/mass spectrometry, and a closed intestinal
An unexpected effect of TNF-alpha on F508del-CFTR maturation and function
Bitam S, et al.
F1000Research, 4 (2015)
Defective CFTR expression and function are detectable in blood monocytes: development of a new blood test for cystic fibrosis
Sorio C, et al.
Testing, 6(7), e22212-e22212 (2011)
Electrophysiological evaluation of cystic fibrosis conductance transmembrane regulator (CFTR) expression in human monocytes
Ettorre M, et al.
Biochimica et biophysica acta. General subjects, 1840(10), 3088-3095 (2014)
Tonghui Ma et al.
The Journal of clinical investigation, 110(11), 1651-1658 (2002-12-05)
Secretory diarrhea is the leading cause of infant death in developing countries and a major cause of morbidity in adults. The cystic fibrosis transmembrane conductance regulator (CFTR) protein is required for fluid secretion in the intestine and airways and, when

Questions

  1. Bonjour,  J'aurai voulu connaitre la stabilité de l'inhibiteur après plusieurs mois après qu'elle soit stocké sous forme de solvant. Merci

    1 answer
    1. It is recommended to aliquot and store frozen at -20°C following reconstitution. Stock solutions are stable for up to 3 months at -20°C.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service