Skip to Content
Merck
All Photos(3)

Key Documents

C2882

Sigma-Aldrich

5β-Cholestan-3α-ol

≥95%

Synonym(s):

Epicoprostanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H48O
CAS Number:
Molecular Weight:
388.67
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.28

Quality Level

Assay

≥95%

form

powder

shipped in

ambient

storage temp.

room temp

SMILES string

CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C27H48O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h18-25,28H,6-17H2,1-5H3/t19-,20-,21-,22+,23-,24+,25+,26+,27-/m1/s1

InChI key

QYIXCDOBOSTCEI-KKFSNPNRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5β-Cholestan-3α-ol was used as internal standard for analysis of sterols in plasma by GC and HPLC.

Biochem/physiol Actions

5β-Cholestan-3α-ol is a sterol produced endogenously from cholesterol and has been isolated from feces of human and rats. It is derived from cholesterol by the action of intestinal microorganisms.

Preparation Note

5β-Cholestan-3α-ol yields clear, colorless to faint yellow solution in chloroform at 50 mg/ml.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative analysis of sterols in serum by high-performance liquid chromatography: application to the biochemical diagnosis of cerebrotendinous xanthomatosis
Kasama T et al
Journal of Chromatography A, 400, 241-246 (1987)
Vikaskumar G Shah et al.
Water research, 41(16), 3691-3700 (2007-07-07)
Faecal samples from humans, herbivores, carnivores and birds as well as samples from septic tanks and effluents from a sewage treatment plant (STP) were extracted and analyzed by gas chromatography-mass spectrometry for faecal sterols including coprostanol, epicoprostanol, cholestanol, cholesterol, stigmasterol
Morgane Derrien et al.
Journal of agricultural and food chemistry, 59(13), 7294-7302 (2011-05-25)
Thirty-five samples of cow feces (cowpat and cow manure) and pig slurries subjected to different treatment processes and different storage times before land spreading were extracted and analyzed by gas chromatography-mass spectrometry to determine their fecal stanol profiles. The fresh
M Souidi et al.
Life sciences, 64(17), 1585-1593 (1999-06-03)
Our purpose was to examine the in vitro modulation of liver mitochondrial sterol 27-hydroxylase (S27OHase) and microsomal cholesterol 7alpha-hydroxylase (CH7alphaOHase) activities by certain drugs, sterols, oxysterols and bile acids, and to compare the influence of sex, age, diet and cholestyramine
Anna N Bukiya et al.
The Journal of general physiology, 137(1), 93-110 (2010-12-15)
The activity (Po) of large-conductance voltage/Ca(2+)-gated K(+) (BK) channels is blunted by cholesterol levels within the range found in natural membranes. We probed BK channel-forming α (cbv1) subunits in phospholipid bilayers with cholesterol and related monohydroxysterols and performed computational dynamics

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service