Skip to Content
Merck
All Photos(1)

Key Documents

C-27020

Sigma-Aldrich

Osteoblast Mineralization Medium

Ready-to-use kit including Basal Medium and SupplementMix, 100 ml

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.81

form

liquid

technique(s)

cell culture | mammalian: suitable

shipped in

wet ice

General description

Osteoblast Mineralization Medium (Ready-to-use)

Application

Osteoblast Mineralization Medium has been developed for the efficient induction of strong mineralization in human osteoblast cultures. Extensive extracellular calcium-deposits can be observed after 21 days by following an easy one-step protocol. The medium is optimized for primary human cells, but can also be used for bovine, murine, and rat osteoblasts as well as osteoblast cell lines.Recommended for:Human Osteoblasts (HOB)

Quality

All lots of PromoCell Osteoblast Mineralization Medium are tested for their capacity to induce mineralization in primary human osteoblasts. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Preparation Note

Contains all the growth factors and supplements necessary for mineralization of human osteoblasts.

Storage and Stability

Store the Basal Medium at 4 to 8°C in the dark and the SupplementMix at -20°C immediately after arrival. Do not freeze the Basal Medium. If stored properly, the products are stable until the expiry date stated on the label. After adding the SupplementMix to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, prewarm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C.

Subculture Routine

Click here for more information.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service