Skip to Content
Merck
All Photos(1)

Key Documents

417-500

Sigma-Aldrich

Human Osteoblast Growth Medium (500ml)

Synonym(s):

Primary Cell Culture Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

form

liquid

Quality Level

shelf life

6 mo.

manufacturer/tradename

Cell Applications, Inc

shipped in

wet ice

storage temp.

2-8°C

General description

All-in-one ready-to-use media

Application

Human Osteoblast Growth Medium (500ml) has been used to culture primary human osteoblasts.

Quality

Each lot was tested for the ability to support plating, spreading and proliferation

Preparation Note

contained in protocol

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

M S Hasan et al.
Journal of the mechanical behavior of biomedical materials, 28, 1-14 (2013-08-21)
Completely resorbable composites are an attractive alternative for metallic bone-fracture fixation devices. However, failure of their interfacial integrity within aqueous environments, which can lead to a rapid loss of overall mechanical properties, has been reported in the literature. In this
Convex vs concave surface nano-curvature of Ta2O5 thin films for tailoring the osteoblast adhesion
Surface and Coatings Technology, 393, 125805-125805 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service