C-24305
Melanocyte Growth Medium M2
Ready-to-use kit including phenol red-free Basal Medium and SupplementMix, 500 ml
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
aqueous solution
packaging
pkg of 500 ml
technique(s)
cell culture | mammalian: suitable
shipped in
wet ice
storage temp.
2-8°C
General description
Melanocyte Growth Medium M2 (Ready-to-use), phenol red-free
Application
Melanocyte Growth Medium M2 is a serum-free and PMA (Phorbol Myristate Acetate)-free Medium that was developed for the optimized in vitro cultivation of human melanocytes. It is also suitable for human pigment epithelial cells, neuronal cells, and melanoma cells. The medium is optimized for primary human cells, but can also be used for equine, porcine, mouse, and rat melanocytes.Recommended for:Normal Human Epidermal Melanocytes (NHEM), juvenile foreskin, cultured in M2 MediumNormal Human Epidermal Melanocytes (NHEM), adult donor, cultured in M2 Medium
Components
Part of a Cell Culture System offered in a variety of formats:Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMixGrowth Medium (Ready-to-use): Includes phenol red-free Basal Medium and SupplementMixSupplementMix: Contains all media supplements premixed in one vial
Quality
All lots of PromoCell Melanocyte Growth Medium M2 are subjected to comprehensive quality control tests using primary human melanocytes. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested melanocytes. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).
Storage and Stability
Store the Basal Medium at 4 to 8°C in the dark and the SupplementMix at -20°C immediately after arrival. Do not freeze the Basal Medium. If stored properly, the products are stable until the expiry date stated on the label. After adding the SupplementMix to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C.
Subculture Routine
Click here for more information.
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service