Skip to Content
Merck
All Photos(1)

Documents

C1231

SAFC

Cholesterol, Plant-Derived

SyntheChol®

Synonym(s):

Cholesterol, Synthetic cholesterol, 3β-Hydroxy-5-cholestene, 5-Cholesten-3β-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46O
CAS Number:
Molecular Weight:
386.65
Beilstein:
1915888
EC Number:
MDL number:
UNSPSC Code:
12352211

biological source

synthetic

Agency

JP
NF
Ph. Eur.

Assay

≥98%

form

crystals

bp

360 °C (lit.)

mp

147-149 °C (lit.)

density

1.067 g/mL at 25 °C (lit.)

suitability

suitable for manufacturing use

shipped in

dry ice

storage temp.

−20°C (−15°C to −25°C)

SMILES string

CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1

InChI key

HVYWMOMLDIMFJA-DPAQBDIFSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Major component of all biological membranes; ~25% of total brain lipid is cholesterol.

Preparation Note

Synthesized from material of non-animal origin

Legal Information

SyntheChol is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steven A Grover et al.
The lancet. Diabetes & endocrinology, 3(2), 114-122 (2014-12-09)
Despite the increased risk of cardiovascular disease and type 2 diabetes associated with excess bodyweight, development of a clinically meaningful metric for health professionals remains a challenge. We estimated the years of life lost and the life-years lost from diabetes
Bo Angelin et al.
Journal of internal medicine, 277(3), 331-342 (2014-04-24)
Liver-selective thyromimetic agents could provide a new approach for treating dyslipidaemia. We performed a multicentre, randomized, placebo-controlled, double-blind study to evaluate the efficacy and safety of eprotirome, a liver-selective thyroid hormone receptor agonist, in 98 patients with primary hypercholesterolaemia. After
Michael J Pencina et al.
The New England journal of medicine, 370(15), 1422-1431 (2014-03-22)
The 2013 guidelines of the American College of Cardiology and the American Heart Association (ACC-AHA) for the treatment of cholesterol expand the indications for statin therapy for the prevention of cardiovascular disease. Using data from the National Health and Nutrition
Matthew P Buman et al.
American journal of epidemiology, 179(3), 323-334 (2013-12-10)
Sleep and sedentary and active behaviors are linked to cardiovascular disease risk biomarkers, and across a 24-hour day, increasing time in 1 behavior requires decreasing time in another. We explored associations of reallocating time to sleep, sedentary behavior, or active
Peter Libby et al.
Immunity, 38(6), 1092-1104 (2013-07-03)
According to the traditional view, atherosclerosis results from a passive buildup of cholesterol in the artery wall. Yet, burgeoning evidence implicates inflammation and immune effector mechanisms in the pathogenesis of this disease. Both innate and adaptive immunity operate during atherogenesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service