B8781
Restriction Enzyme Buffer SB
10 ×
Synonym(s):
Restriction Endonuclease Buffer
About This Item
Recommended Products
grade
for molecular biology
Quality Level
description
10× buffer contains 100 mM Tris-HCl, pH 8.0, 50 mM MgCl2, 1000 mM NaCl, 10 mM 2-mercaptoethanol at 37 °C.
form
buffered aqueous solution
concentration
10 ×
storage temp.
−20°C
SMILES string
CC(C)CC(CNC(Cc1ccccc1)C(N)=O)NC(=O)C(Cc2cnc[nH]2)NC(=O)CNC(=O)C(NC(=O)C(C)NC(=O)C(Cc3c[nH]c4ccccc34)NC(=O)C(CCC(N)=O)NC(=O)C(N)Cc5c[nH]c6ccccc56)C(C)C
InChI
1S/C58H77N15O9/c1-32(2)21-39(29-65-46(52(61)76)22-35-13-7-6-8-14-35)69-57(81)48(25-38-28-62-31-67-38)70-50(75)30-66-58(82)51(33(3)4)73-53(77)34(5)68-56(80)47(24-37-27-64-44-18-12-10-16-41(37)44)72-55(79)45(19-20-49(60)74)71-54(78)42(59)23-36-26-63-43-17-11-9-15-40(36)43/h6-18,26-28,31-34,39,42,45-48,51,63-65H,19-25,29-30,59H2,1-5H3,(H2,60,74)(H2,61,76)(H,62,67)(H,66,82)(H,68,80)(H,69,81)(H,70,75)(H,71,78)(H,72,79)(H,73,77)
InChI key
YNZJHLFHFNCZIE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service