Skip to Content
Merck
All Photos(1)

Documents

B7435

Sigma-Aldrich

CelLytic B Cell Lysis Reagent

For bacterial cell lysis, standard strength

Synonym(s):

Cell lysis reagent

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116134
NACRES:
NA.77

Quality Level

form

solution

Looking for similar products? Visit Product Comparison Guide

Application

CelLytic B Cell Lysis Reagent has been used for cell lysis for the extraction of coenzyme Q10, recombinant Src kinase associated phosphoprotein 1- glutathione S-transferases (GST) fusion protein and human α-synuclein.
A proprietary, non-denaturing formulation of zwitterionic detergents used for the lysis of bacterial cells and extraction of recombinant proteins.

Features and Benefits

  • Higher protein extraction efficiency than traditional methods such as sonication and lysozyme
  • Scalable for 1 to 25 grams of bacterial cell paste
  • No interference with downstream applications such as affinity chromatography, IP, and Western blotting
  • Compatible with protease inhibitors, inhibitor cocktails, chaotropes, salts, chelating agents and reducing agents

Other Notes

Detergent blend formulated in 40 mM Trizma® HCl (pH 8.0).

Legal Information

Covered by US Patent No 7,282,475 B2 and are sold for research use only. Commercial use requires addtional licenses.
CelLytic is a trademark of Sigma-Aldrich Co. LLC
Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ken Motohashi
BMC biotechnology, 15, 47-47 (2015-06-04)
Seamless ligation cloning extract (SLiCE) is a simple and efficient method for DNA assembly that uses cell extracts from the Escherichia coli PPY strain, which expresses the components of the λ prophage Red/ET recombination system. This method facilitates restriction endonuclease
Ca2+ increases the specific coenzyme Q10 content in Agrobacterium tumefaciens
Ha SJ, et al.
Bioprocess and Biosystems Engineering, 32(5), 697-700 (2009)
Prion-like seeding of misfolded alpha-synuclein in the brains of dementia with Lewy body patients in RT-QUIC
Sano K, et al.
Molecular Neurobiology, 55(5), 3916-3930 (2018)
Kazunori Sano et al.
Acta neuropathologica communications, 9(1), 182-182 (2021-11-14)
Serine 129 (S129) phosphorylation of α-synuclein (αSyn) is a central feature of Lewy body (LB) disease pathology. Although the neighboring tyrosine residues Y125, Y133, and Y136 are also phosphorylation sites, little is known regarding potential roles of phosphorylation cross-talk between
Immune adaptor protein SKAP1 (SKAP-55) forms homodimers as mediated by the N-terminal region
Raab M, et al.
BMC Research Notes, 11(1), 869-869 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service