Skip to Content
Merck
All Photos(3)

Documents

B6396

Sigma-Aldrich

2′(3′)-O-(4-Benzoylbenzoyl)adenosine 5′-triphosphate triethylammonium salt

≥93%

Synonym(s):

Benzoylbenzoyl-ATP, Bz-ATP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H24N5O15P3 · xC6H15N × yH2O
CAS Number:
Molecular Weight:
715.39 (anhydrous free acid basis)
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

Assay

≥93%

form

powder

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

CCN(CC)CC.Nc1ncnc2n(cnc12)[C@@H]3O[C@H](COP(O)(=O)OP(O)(=O)OP(O)(O)=O)[C@@H](OC(=O)c4ccc(cc4)C(=O)c5ccccc5)[C@H]3O

InChI

1S/C24H24N5O15P3.C6H15N/c25-21-17-22(27-11-26-21)29(12-28-17)23-19(31)20(16(41-23)10-40-46(36,37)44-47(38,39)43-45(33,34)35)42-24(32)15-8-6-14(7-9-15)18(30)13-4-2-1-3-5-13;1-4-7(5-2)6-3/h1-9,11-12,16,19-20,23,31H,10H2,(H,36,37)(H,38,39)(H2,25,26,27)(H2,33,34,35);4-6H2,1-3H3/t16-,19-,20-,23-;/m1./s1

InChI key

HVOVBTNCGADRTH-WBLDMZOZSA-N

General description

2′(3′)-O-(4-Benzoylbenzoyl)adenosine 5′-triphosphate is a photoreactive analog of ATP.

Application

2′(3′)-O-(4-Benzoylbenzoyl)adenosine 5′-triphosphate triethylammonium salt has been used as a P2X4 receptor agonist in rat kidney sections, type I diabetes induced rats and astrocyte−neuron coculture. It has also been used as a synthetic ATP analog in isotonic E3 medium.
Useful as a photoaffinity label for proteins and enzymes that bind ATP.

Biochem/physiol Actions

2′(3′)-O-(4-Benzoylbenzoyl)adenosine 5′-triphosphate triethylammonium salt is a selective purinergic receptor (P2X) agonist. It is more potent than ATP at homodimeric P2X7 receptors. It also effectively activates P2X1, P2X2, and P2X3 receptors. Useful as a photoaffinity label for proteins and enzymes that bind ATP.
Selective P2X purinergic agonist. It is more potent than ATP at homodimeric P2X7 receptors.

Other Notes

Mixed isomers

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan Francisco Codocedo et al.
PloS one, 8(3), e57626-e57626 (2013-03-09)
To assess the putative role of adenosine triphosphate (ATP) upon nitric oxide (NO) production in the hippocampus, we used as a model both rat hippocampal slices and isolated hippocampal neurons in culture, lacking glial cells. In hippocampal slices, additions of
Covalent binding of 3'-O-(4-benzoyl) benzoyl adenosine 5'-triphosphate (BzATP) to the isolated alpha and beta subunits and the alpha 3 beta 3 core complex of TF1. Covalent binding of BzATP prevents association of alpha and beta subunits and induces dissociation of the alpha 3 beta 3 core complex.
BarZvi D, et al.
The Journal of Biological Chemistry, 267(16), 11029-11033 (1992)
Diabetes-induced Neuropathic Mechanical Hyperalgesia Depends on P2X4 Receptor Activation in Dorsal Root Ganglia
Teixeira JM, et al.
Neuroscience, 398, 158-170 (2019)
Ji Zhao et al.
Molecular neurodegeneration, 4, 4-4 (2009-01-16)
Insulin-degrading enzyme (IDE) is a ubiquitously expressed zinc-metalloprotease that degrades several pathophysiologically significant extracellular substrates, including insulin and the amyloid beta-protein (Abeta), and accumulating evidence suggests that IDE dysfunction may be operative in both type 2 diabetes mellitus and Alzheimer
Molecular and functional properties of P2X receptors?recent progress and persisting challenges
Kaczmarek-Hajek K, et al.
Purinergic Signaling, 8(3), 375-417 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service