Skip to Content
Merck
All Photos(2)

Key Documents

B3773

Sigma-Aldrich

Anti-Human IgG (Fc specific)−Biotin antibody, Mouse monoclonal

clone HP-6017, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-Human IgG (Fc specific)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

conjugate

biotin conjugate

antibody form

purified immunoglobulin

antibody product type

secondary antibodies

clone

HP-6017, monoclonal

form

buffered aqueous solution

species reactivity

rabbit, sheep, horse (IgG), goat, human

technique(s)

direct ELISA: 1:80,000

isotype

IgG2a

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Immunoglobulin IgG is an abundant protein in human serum. The four classes of IgG include IgG1, IgG2, IgG3 and IgG4.. The IgG heavy chain region is mapped to human chromosome 14. Immunoglobulin super-family have a common structure, comprising of two heavy (H) chains and two light (L) chains, held together by disulfide linkages. The heavy chain has one variable N-terminal region and three to four constant (CH1-CH4) C-terminal regions. The L chain comprises of one variable N-terminal region and a constant C-terminal region.

Specificity

The antibody is specific for the Fc portion of human IgG and recognizes an epitope common to all human IgG subclasses. This antibody was adopted as an Fc specific reagent in the IUIS/WHO study.

Application

Anti-Human IgG (Fc specific) Biotin antibody, Mouse monoclonal has been used in fluoroimmunoassay of plasma samples.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Enzyme-linked immunosorbent assay (1 paper)
Monoclonal Anti-Human IgG (Fc specific)-Biotin antibody produced in mouse is suitable for ELISA at a working dilution of 1:80,000. It was used for labeling mouse splenocytes and analysis by flow cytometry.

Biochem/physiol Actions

IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections. The coupling of biotin to monoclonal Anti-Human IgG (Fc specific) antibody allows for the binding of various labels such as avidin or streptavidin.
IgG1 class is the most abundant and its deficiency results in hypogammaglobulinemia. IgG2 deficiency increases susceptibility to bacterial infections. IgG3 mediates effector functions, and IgG4 is associated with asymptomatic infection. Digestion of IgG by papain results in the generation of fragment antigen binding (Fab). Pepsin digestion of IgG generates fragment crystallisable (Fc). The Fc region of IgG antibody has enormous therapeutic potential and is exploited for the development of therapeutic antibodies.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intracellular cytokines may model immunoregulation of abacavir hypersensitivity in HIV-infected subjects.
King D, et al.
The Journal of Allergy and Clinical Immunology, 115(5), 1081-1087 (2005)
C Whitney et al.
Infection and immunity, 60(6), 2194-2200 (1992-06-01)
Porphyromonas gingivalis is a suspected pathogen in rapidly progressive periodontitis (RPP). We have determined the anti-P. gingivalis serum immunoglobulin G (IgG) isotype response and avidity and the subclass titer distributions for 30 RPP patients and 30 age-, sex-, and race-matched
Gunnveig Grødeland et al.
PloS one, 8(11), e80008-e80008 (2013-11-19)
Different diseases require different immune responses for efficient protection. Thus, prophylactic vaccines should prime the immune system for the particular type of response needed for protection against a given infectious agent. We have here tested fusion DNA vaccines which encode
María Eugenia Toledo-Romani et al.
Med (New York, N.Y.) (2022-08-24)
SOBERANA 02 has been evaluated in phase I and IIa studies comparing homologous versus heterologous schedule (this one, including SOBERANA Plus). Here, we report results of immunogenicity, safety, and reactogenicity of SOBERANA 02 in a two- or three-dose heterologous scheme
Divya Thiagarajan et al.
Journal of immunology (Baltimore, Md. : 1950), 205(8), 2109-2116 (2020-09-06)
Abs against phosphorylcholine (anti-PC) and Abs against malondialdehyde (anti-MDA) may be protective in chronic inflammation, like atherosclerosis and cardiovascular disease. It is not known how they develop early in life. Ab titers were measured using ELISA in healthy women (n

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service