Skip to Content
Merck
All Photos(1)

Key Documents

B3523

Sigma-Aldrich

Anti-Human IgG3−Biotin antibody, Mouse monoclonal

clone HP-6050, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-Human IgG3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

mouse

conjugate

biotin conjugate

antibody form

purified from hybridoma cell culture

antibody product type

secondary antibodies

clone

HP-6050, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

direct ELISA: 1:40,000

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Human IgG consists of four subclasses (1-4) that can be recognized by antigenic differences in their heavy chains. They constitute approximately 65, 30, 5 and 4% of the total IgG, respectively. Each subclass has different biological and physicochemical properties. The IgG subclass may be preferentially produced in response to different antigens and pathological conditions. For instance, anti-polysaccharide responses are mainly of the IgG2 subclass while protein antigens give rise to IgG1 and IgG3 antibodies.
Monoclonal Anti-Human IgG3 (mouse IgG1 isotype) is derived from the hybridoma (HP-60501) produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse.

Specificity

The antibody is specific for human IgG3 and is non-reactive with other IgG subclasses. This clone has been recommended by the IUIS/WHO study for its specificity and reactivity in a wide range of assay protocols and techniques.

Immunogen

Purified myeloma human IgG3 coupled to polyaminostyrene (PAS) microbeads was used as the immunogen

Application

The product may be used for the identification of human IgG3 subclass by means of various immunoassays and ELISA. It has also been used in immunoblotting.

Biochem/physiol Actions

IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections. The coupling of biotin to monoclonal Anti-Human IgG3 antibody allows for the binding of various labels such as avidin or streptavidin.
Only IgG1 and IgG3 are capable of adherence to mononuclear phagocytes. IgG3 has O-glycosylation sites and mediates, phagocytosis, antibody-mediated cellular cytotoxicity, neutralization, and activates complement pathways. IgG3 subtypes have short in vivo half-life and mediates the effector functions than IgG1.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing, or storage in "frost-free" freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carolin Cornelius et al.
EBioMedicine, 11, 58-67 (2016-08-29)
We have constructed and clinically evaluated a hypoallergenic vaccine for grass pollen allergy, BM32, which is based on fusion proteins consisting of peptides from the IgE binding sites of the major grass pollen allergens fused to preS (preS1+preS2), a domain
A unique phenotype of acquired Glanzmann thrombasthenia due to non-function-blocking anti-?IIb?3 autoantibodies
Akuta, K, et al.
Journal of Thrombosis and Haemostasis, 17(1), 206-219 (2019)
Role of IgG3 in Infectious Diseases
Damelang T, et al.
Trends in Immunology, 40(3), 197-211 (2019)
Torunn Elisabeth Tjelle et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 9(3), 328-336 (2004-03-10)
Antibodies are useful for the treatment of a variety of diseases. We here demonstrate that mouse muscle produced monoclonal antibodies (mAb) after a single injection of immunoglobulin genes as plasmid DNA. In vivo electroporation of muscle greatly enhanced antibody production.
Anti-contactin-associated protein-2 encephalitis: relevance of antibody titres, presentation and outcome
Bien, CG, et al.
European Journal of Neurology, 24(1), 175-186 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service