Skip to Content
Merck
All Photos(2)

Key Documents

AV48136

Sigma-Aldrich

Anti-PDHA1 (AB1) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-PDHA, Anti-PDHCE1A, Anti-PHE1A, Anti-Pyruvate dehydrogenase (lipoamide) α 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

40 kDa

species reactivity

human, mouse, guinea pig, rabbit, dog, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PDHA1(5160)

General description

PDHA1 codes for the α1 subunit of the pyruvate dehydrogenase complex and modulates the activity of the complex. PDHA1 mutations have been implicated in pyruvate dehydrogenase deficiency.
Rabbit Anti-PDHA1 antibody recognizes bovine, chicken, pig, human, mouse, rat, and canine PDHA1.

Immunogen

Synthetic peptide directed towards the N terminal region of human PDHA1

Application

Rabbit Anti-PDHA1 antibody is suitable for western blot applications at a concentration of 5μg/ml.

Biochem/physiol Actions

The pyruvate dehydrogenase complex is a nuclear-encoded mitochondrial matrix multienzyme complex that provides the primary link between glycolysis and the tricarboxylic acid (TCA) cycle by catalyzing the irreversible conversion of pyruvate into acetyl-CoA. The PDH complex is composed of multiple copies of 3 enzymes: E1 (PDHA1); dihydrolipoyl transacetylase (DLAT); and dihydrolipoyl dehydrogenase (DLD). The E1 enzyme is a heterotetramer of 2 alpha and 2 beta subunits. The E1-alpha subunit contains the E1 active site and plays a key role in the function of the PDH complex.The pyruvate dehydrogenase complex is a nuclear-encoded mitochondrial matrix multienzyme complex that provides the primary link between glycolysis and the tricarboxylic acid (TCA) cycle by catalyzing the irreversible conversion of pyruvate into acetyl-CoA. The PDH complex is composed of multiple copies of 3 enzymes: E1 (PDHA1); dihydrolipoyl transacetylase (DLAT; MIM 608770) (E2; EC 2.3.1.12); and dihydrolipoyl dehydrogenase (DLD; MIM 238331) (E3; EC 1.8.1.4). The E1 enzyme is a heterotetramer of 2 alpha and 2 beta subunits. The E1-alpha subunit contains the E1 active site and plays a key role in the function of the PDH complex (Brown et al., 1994 [PubMed 7853374]).[supplied by OMIM]. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Sequence

Synthetic peptide located within the following region: MRKMLAAVSRVLSGASQKPASRVLVASRNFANDATFEIKKCDLHRLEEGP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Ostergaard et al.
Journal of inherited metabolic disease, 32 Suppl 1, S235-S239 (2009-06-12)
The pyruvate dehydrogenase (PDH) complex is a mitochondrial multienzyme that catalyses the irreversible oxidative decarboxylation of pyruvate to acetyl-CoA. We report four novel PDHA1 mutations in patients with pyruvate dehydrogenase deficiency. Analysis of PDH activity showed decreased activity in fibroblasts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service