Skip to Content
Merck
All Photos(1)

Key Documents

AP1063

Sigma-Aldrich

PhosphoDetect Anti-PDH-E1α (pSer²³²) Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-Pyruvate Dehydogenase pSer²³² Rabbit pAb

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

mouse, human, rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer232)

General description

Immunoaffinity purified rabbit polyclonal antibody. Recognizes the ~44 kDa PDH-E1α protein phosphorylated at Ser232.
Recognizes the ~44 kDa PDH-E1α protein phosphorylated at Ser232 in HEK293 cells.
This PhosphoDetect Anti-PDH-E1α (pSer²³²) Rabbit pAb is validated for use in Immunoblotting, IF, IP for the detection of PDH-E1α (pSer²³²).

Immunogen

Human
a synthetic phosphopeptide corresponding to amino acids surrounding the Ser²³² phosphorylation site of human PDH-E1α

Application



Immunoblotting (0.25 g/ml)
Immunofluorescence (Please see comments)
Immunoprecipitation (1 g/ml)

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
HEK293 cells

Other Notes

Rardin M.J., et. al. 2009. Anal. Biochem.2, 157.
Seifert, F., et al. 2007. Biochemistry 21, 6277.
Lee, J., et al. 2007. Mol. Cell Prot. 4, 669.
Patel, M.S. and Korotchkina, L.G. 2006 Biochem. Soc. Trans.34, 217.
Korotchkina, L.G., et al. 2001. J. Biol. Chem. 40, 37223.
This antibody has been known to work with immunofluorescence. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lærke Bertholdt et al.
Journal of applied physiology (Bethesda, Md. : 1985), 124(3), 729-740 (2017-12-02)
Recruitment of fatty acids from adipose tissue is increased during fasting. However, the molecular mechanisms behind fasting-induced metabolic regulation in human adipose tissue and the potential impact of training state in this are unknown. Therefore the aim of the present
Shelby L Oke et al.
Reproduction (Cambridge, England), 159(1), 27-39 (2019-11-07)
Epidemiological data suggest an inverse relationship between birth weight and long-term metabolic deficits, which is exacerbated by postnatal catch-up growth. We have previously demonstrated that rat offspring subject to maternal protein restriction (MPR) followed by catch-up growth exhibit impaired hepatic
Lærke Bertholdt et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 312(4), R626-R636 (2017-01-27)
The liver is essential in maintaining and regulating glucose homeostasis during prolonged exercise. IL-6 has been shown to be secreted from skeletal muscle during exercise and has been suggested to signal to the liver. Therefore, the aim of this study
Arpit Sharma et al.
eLife, 8 (2019-07-16)
Metabolic cycles are a fundamental element of cellular and organismal function. Among the most critical in higher organisms is the Cori Cycle, the systemic cycling between lactate and glucose. Here, skeletal muscle-specific Mitochondrial Pyruvate Carrier (MPC) deletion in mice diverted
Rosa Ferriero et al.
Science translational medicine, 5(175), 175ra31-175ra31 (2013-03-08)
Lactic acidosis is a buildup of lactic acid in the blood and tissues, which can be due to several inborn errors of metabolism as well as nongenetic conditions. Deficiency of pyruvate dehydrogenase complex (PDHC) is the most common genetic disorder

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service