Skip to Content
Merck
All Photos(3)

Key Documents

AV46114

Sigma-Aldrich

Anti-APOBEC3B antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-APOBEC1L, Anti-ARCD3, Anti-ARP4, Anti-Apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3B, Anti-DJ742C19.2, Anti-FLJ21201, Anti-PHRBNL, Anti-bK150C2.2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

46 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... APOBEC3B(9582)

Immunogen

Synthetic peptide directed towards the N terminal region of human APOBEC3B

Application

Anti-APOBEC3B antibody produced in rabbit is suitable for western blotting at a concentration of 0.25μg/ml.

Biochem/physiol Actions

APOBEC3B is a member of the cytidine deaminase gene family and encodes apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3B. Endogenous APOBEC3B is a principal nuclear protein that facilitates the DNA C-to-U editing activity in breast cancer cell-line extracts. APOBEC3B exhibits antiviral activity against human T-cell leukemia virus type 1 (HTLV-1) and is a potent inhibitor of simian immunodeficiency virus (SIV) replication.

Sequence

Synthetic peptide located within the following region: NQLPAYKCFQITWFVSWTPCPDCVAKLAEFLSEHPNVTLTISAARLYYYW

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qin Yu et al.
The Journal of biological chemistry, 279(51), 53379-53386 (2004-10-07)
In the human genome the apolipoprotein B mRNA-editing enzyme catalytic polypeptide (APOBEC)3 gene has expanded into a tandem array of genes termed APOBEC3A-G. Two members of this family, APOBEC3G and APOBEC3F, have been found to have potent activity against virion
Marcel Ooms et al.
Journal of virology, 86(11), 6097-6108 (2012-03-30)
The human APOBEC3 family consists of seven cytidine deaminases (A3A to A3H), some of which display potent antiretroviral activity against HIV-1 and other retroviruses. Studies that analyzed the effect of A3G on human T-lymphotropic virus type 1 (HTLV-1) infectivity resulted
Michael B Burns et al.
Nature, 494(7437), 366-370 (2013-02-08)
Several mutations are required for cancer development, and genome sequencing has revealed that many cancers, including breast cancer, have somatic mutation spectra dominated by C-to-T transitions. Most of these mutations occur at hydrolytically disfavoured non-methylated cytosines throughout the genome, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service