Skip to Content
Merck
All Photos(3)

Key Documents

AV39465

Sigma-Aldrich

Anti-NR2F2 (AB2) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-ARP1, Anti-COUP-TFII, Anti-COUPTFB, Anti-MGC117452, Anti-Nuclear receptor subfamily 2, group F, member 2, Anti-SVP40, Anti-TFCOUP2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

45 kDa

species reactivity

guinea pig, human, bovine, dog, rat, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NR2F2(7026)

General description

NR2F2 codes for a steroid hormone receptor that is involved in the differentiation of human stem cells and trophoblasts. Genetic variations in NR2F2 are associated with congenital cardiac disorders.
Rabbit Anti-NR2F2 antibody recognizes human, mouse, rat, zebrafish, canine, bovine, and chicken NR2F2.

Immunogen

Synthetic peptide directed towards the N terminal region of human NR2F2

Application

Rabbit Anti-NR2F2 antibody is suitable for western blot (2.5 μg/ml) and IHC (4-8 μg/ml) applications.

Biochem/physiol Actions

NR2F2, an orphan member of the nuclear hormone receptor superfamily, acts as a transcriptional repressor by antagonizing the functions of other nuclear hormone receptors and by actively silencing transcription. However, in certain contexts, NR2F2 stimulates transcription directly. NR2F2, MyoD and p300 interact in a competitive manner, and that increasing amounts of NR2F2 have the ability to reduce the interaction between myoD and p300 invitro. NR2F2 post-transcriptionally regulates myoD activity/function, and that crosstalk between orphan nuclear receptors and the myogenic bHLH proteins has functional consequences for differentiation.

Sequence

Synthetic peptide located within the following region: QDEVPGSQGSQASQAPPVPGPPPGAPHTPQTPGQGGPASTPAQTAAGGQG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saeed Al Turki et al.
American journal of human genetics, 94(4), 574-585 (2014-04-08)
Congenital heart defects (CHDs) are the most common birth defect worldwide and are a leading cause of neonatal mortality. Nonsyndromic atrioventricular septal defects (AVSDs) are an important subtype of CHDs for which the genetic architecture is poorly understood. We performed
Daniela C Dos Santos et al.
Oxidative medicine and cellular longevity, 2022, 3710449-3710449 (2022-04-02)
Glioblastoma multiforme (GBM) is a notably lethal brain tumor associated with high proliferation rate and therapeutic resistance, while currently effective treatment options are still lacking. Imidazo[1,2-a]pyridine derivatives and organoselenium compounds are largely used in medicinal chemistry and drug development. This
Michael A Hubert et al.
PloS one, 5(2), e9417-e9417 (2010-03-03)
During the in vitro differentiation of human villous cytotrophoblast (CTB) cells to a syncytiotrophoblast (STB) phenotype, mRNA levels for the nuclear hormone receptor NR2F2 (ARP-1, COUP-TFII) increase rapidly, reaching a peak at day 1 of differentiation that is 8.8-fold greater
Alessandro Rosa et al.
The EMBO journal, 30(2), 237-248 (2010-12-15)
Multiple levels of control are in play to regulate pluripotency and differentiation in human embryonic stem cells (hESCs). At the transcriptional level, the core factors OCT4, NANOG and SOX2 form a positive autoregulatory loop that is pivotal for maintaining the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service