Skip to Content
Merck
All Photos(1)

Key Documents

A9861

Sigma-Aldrich

Auraptene

≥98% (HPLC)

Synonym(s):

7-Geranyloxycoumarin, 7-[[(2E)-3,7-dimethyl-2,6-octadien-1-yl]oxy]-2H-1-benzopyran-2-one, Aurapten

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22O3
CAS Number:
Molecular Weight:
298.38
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >20 mg/mL

storage temp.

−20°C

SMILES string

C\C(C)=C\CC\C(C)=C\COc1ccc2C=CC(=O)Oc2c1

InChI

1S/C19H22O3/c1-14(2)5-4-6-15(3)11-12-21-17-9-7-16-8-10-19(20)22-18(16)13-17/h5,7-11,13H,4,6,12H2,1-3H3/b15-11+

InChI key

RSDDHGSKLOSQFK-RVDMUPIBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Auraptene (7-geranyloxycoumarin) is a prenyloxycoumarin, that is abundantly present in nature. It is produced by several plant species, such as Rutaceae and Umbeliferae (Apiaceae) families.

Biochem/physiol Actions

Auraptene is an ACAT inhibitor and estrogen receptor modulator.
Auraptene is an estrogen receptor modulator that also acts as an ACAT inhibitor. It displays anti-tumor effects in several xenograft and chemically-induced murine tumor models. Auraptene induces apoptosis and is anti-proliferative in cancer cell lines. It has agonistic properties against PPARs, and interferes with lipid and cholesterol production by inhibiting ACAT (IC50 = 4 uM).
Auraptene (7-geranyloxycoumarin) possesses anti-oxidant, anti-bacterial, anti-inflammatory and anti-tumor activities.

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satoshi Okuyama et al.
European journal of pharmacology, 699(1-3), 118-123 (2012-12-12)
Cerebral ischemia causes delayed neuronal cell death in the hippocampus resulting in sequential cognitive impairments. Hyper-activated inflammation following ischemia is one of the etiologies for delayed neuronal cell death. In the present study, using a transient global ischemia mouse model
Fatemeh Soltani et al.
Phytotherapy research : PTR, 24(1), 85-89 (2009-05-16)
The antigenotoxicity effects of auraptene on DNA damage in human peripheral lymphocytes were studied using alkaline single cell gel electrophoresis. Auraptene at concentrations of 5, 10, 25, 50, 100, 200 and 400 microM was tested under simultaneous treatment with 25
Takuji Tanaka et al.
Nutrition and cancer, 60 Suppl 1, 70-80 (2008-11-15)
Dietary polyphenols are important potential chemopreventive natural agents. Other agents, such as citrus compounds, are also candidates for cancer chemopreventives. They act on multiple key elements in signal transduction pathways related to cellular proliferation, differentiation, apoptosis, inflammation, and obesity. This
Akira Murakami
Forum of nutrition, 61, 193-203 (2009-04-16)
A regulated low level of nitric oxide (NO) production in the body is essential for maintaining homeostasis (neuroprotection, vasorelaxation, etc.), though certain pathophysiological conditions associated with inflammation involve de novo synthesis of inducible NO synthase (iNOS) in immune cells, including
Auraptene and Its Role in Chronic Diseases
Drug Discovery from Mother Nature, 399-407 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service