Skip to Content
Merck
All Photos(1)

Documents

A9469

Sigma-Aldrich

Monoclonal ANTI-FLAG® M2-Alkaline Phosphatase antibody produced in mouse

clone M2, purified immunoglobulin, buffered aqueous glycerol solution

Synonym(s):

Monoclonal ANTI-FLAG® M2 antibody produced in mouse, Anti-ddddk, Anti-dykddddk

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.32

biological source

mouse

conjugate

alkaline phosphatase conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

M2, monoclonal

form

buffered aqueous glycerol solution

species reactivity

all

concentration

~1 mg/mL

technique(s)

indirect ELISA: 1:20,000

isotype

IgG1

immunogen sequence

DYKDDDDK

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ALPL(249)

Looking for similar products? Visit Product Comparison Guide

General description

Monoclonal ANTI-FLAG® M2-Alkaline Phosphatase is a purified IgG 1 mouse antibody covalently conjugated to calf intestinal alkaline phosphatase (AP). The antibody conjugate binds to FLAG® fusion proteins and will recognize the FLAG® epitope at any position in the fusion protein (N-terminal, Met-N-terminal, C-terminal or internal FLAG® peptides).

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)
Monoclonal ANTI-FLAG® M2-Alkaline Phosphatase antibody produced in mouse has been used:
  • in direct tissue blot immunoassay of sweet orange petioles samples
  • in screening internalization of delta opioid receptor
  • for screening cell-free protein expression using ELISA

Physical form

Solution in Tris buffered saline containing 50% glycerol plus stabilizer and preservative

Legal Information

ANTI-FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Waithaka Mwangi et al.
Journal of leukocyte biology, 78(2), 401-411 (2005-04-29)
Induction of immune responses against microbial antigens using DNA is an attractive strategy to mimic the immunity induced by live vaccines. Although DNA vaccines are efficacious in murine models, the requirement for multiple immunizations using high doses in outbred animals
Integration of cell-free protein coexpression with an enzyme-linked immunosorbent assay enables rapid analysis of protein-protein interactions directly from DNA
Layton CJ and Helling HW
Protein Science, 20(8), 1432-1438 (2011)
Beth A Rasala et al.
Plant biotechnology journal, 9(6), 674-683 (2011-05-04)
Microalgae have the potential to be a valuable biotechnological platform for the production of recombinant proteins. However, because of the complex regulatory network that tightly controls chloroplast gene expression, heterologous protein accumulation in a wild-type, photosynthetic-competent algal chloroplast remains low.
Seongjoon Kang et al.
Biology, 7(2) (2018-05-09)
Chlamydomonas reinhardtii (Chlamydomonas) strains that are toxic to mosquito larvae because they express chloroplast transgenes that are based on the mosquitocidal proteins of Bacillus thuringiensis subsp. israelensis (Bti) could be very useful in mosquito control. Chlamydomonas has several advantages for
Úrsula Flores-Pérez et al.
Plant physiology, 170(1), 147-162 (2015-11-21)
The Hsp100-type chaperone Hsp93/ClpC has crucial roles in chloroplast biogenesis. In addition to its role in proteolysis in the stroma, biochemical and genetic evidence led to the hypothesis that this chaperone collaborates with the inner envelope TIC complex to power

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service