Skip to Content
Merck
All Photos(2)

Key Documents

A3682

Sigma-Aldrich

Anti-Mouse IgG (Fab specific)–Peroxidase antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Goat Anti-Mouse IgG (Fab specific)- HRP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse

should not react with

human, rat

technique(s)

direct ELISA: 1:40,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:150
western blot: 1:80,000-160,000 using detecting β-actin in total cell extract of HeLa cells (5-10 μg per lane)

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Immunoglobulin G (IgG) belongs to the immunoglobulin family and is a widely expressed serum antibody. An immunoglobulin has two heavy chains and two light chains connected by a disulfide bond. It mainly helps in immune defense. It is a glycoprotein. IgG is a major class of immunoglobulin. Mice possess five classes of immunoglobulins- IgM, IgG, IgA, IgD and IgE. Mouse IgG is further divided into five classes- IgG1, IgG2a, IgG2b and IgG3.
Immunoglobulin G (IgG) is a glycoprotein antibody that regulates immune responses such as phagocytosis and is also involved in the development of autoimmune diseases. Mouse IgGs have four distinct isotypes, namely, IgG1, IgG2a, IgG2b, and IgG3. IgG1 regulates complement fixation in mice.
Goat anti-mouse IgG (Fab specific)-peroxidase antibody is specific for mouse IgG and the Fab fragment of mouse IgG. The antibody reacts with all mouse IgG subclasses (G1, G2a, G2b and G3) and also with mouse IgA, IgM and IgE. Moreover, the antibody conjugate shows no reactivity with the Fc fragment of mouse IgG or with human and rat IgG.

Immunogen

purified mouse IgG, Fab fragment

Application

Murine IgG levels were quantitated in brain abscess homogenates by ELISA using HRP conjugated goat anti-mouse Fab specific antibody as the detection antibody.
Anti-Mouse IgG (Fab specific)-Peroxidase antibody produced in goat has been used in:
  • enzyme-linked immunosorbent assay (ELISA)
  • immunohistochemical studies
  • dot or immunoblotting

Goat anti-mouse IgG (Fab specific)-peroxidase antibody can be used for western blotting applications. The product can also be used for direct ELISA (1:40,000) and immunohistochemistry (at 1:150, using formalin-fixed, paraffin-embedded sections).
HRP conjugated goat anti-mouse IgG Fab specific was used as the secondary antibody for western blot analysis of bacterial samples of s. equi cells to detect fibrinogen-binding proteins.

Biochem/physiol Actions

Immunoglobulin G (IgG) participates in hypersensitivity type II and type III reactions. IgG2a has cytotropic properties and IgG2 molecules have the ability to activate the complement system. IgG helps in opsonization, complement fixation and antibody dependent cell mediated cytotoxicity.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 0.05% MIT

Preparation Note

Adsorbed with human IgG and rat serum proteins to reduce background staining with human or rat samples.
Prepared by the two-step glutaraldehyde method described by Avrameas, S., et al., Scand. J. Immunol., 8, Suppl. 7, 7 (1978).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quercetin, not caffeine, is a major neuroprotective component in coffee
Lee M, et al.
Neurobiology of Aging, 46, 113-123 (2016)
Acidic fibroblast growth factor (FGF) potentiates glial-mediated neurotoxicity by activating FGFR2 IIIb protein
Lee M, et al.
The Journal of Biological Chemistry, 286(48), 41230-41245 (2011)
M Meehan et al.
Microbiology (Reading, England), 147(Pt 12), 3311-3322 (2001-12-12)
The major cell-wall-associated protein of the equine pathogen Streptococcus equi subsp. equi is an M-like fibrinogen-binding protein (FgBP) which binds equine fibrinogen (Fg) avidly, through residues located at the extreme N-terminus of the molecule. In this study, it is shown
Astrocytes produce the antiinflammatory and neuroprotective agent hydrogen sulfide
Lee M, et al.
Neurobiology of Aging, 30(10), 1523-1534 (2009)
Martin Parizek et al.
BioMed research international, 2013, 371430-371430 (2013-04-16)
The attractiveness of synthetic polymers for cell colonization can be affected by physical, chemical, and biological modification of the polymer surface. In this study, low-density polyethylene (LDPE) was treated by an Ar(+) plasma discharge and then grafted with biologically active

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service