Skip to Content
Merck
All Photos(1)

Key Documents

A8706

Sigma-Aldrich

Avidin from egg white

recombinant, expressed in corn, ≥12 units/mg protein (E1%/280)

Synonym(s):

AVID_CHICK

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.56

recombinant

expressed in corn

Assay

≥80% protein basis (biuret)

form

powder

specific activity

≥12 units/mg protein (E1%/280)

mol wt

glycoprotein 66 kDa
subunit 16 kDa

technique(s)

ELISA: suitable
immunoblotting: suitable
immunoelectrophoresis: suitable

UniProt accession no.

storage temp.

2-8°C

Gene Information

chicken ... AVD(396260)

Looking for similar products? Visit Product Comparison Guide

General description

Avidin is a homotetrameric glycoprotein found in the egg white of birds, reptiles and amphibians. Each subunit is 16 kDa, singly glycosylated and binds to a molecule of biotin with greater affinity and specificity. Recombinant avidin from corn is similar to avidin from egg white in terms of properties like isoelectric point (pI) and antigenic properties except for the molecular weight. Avidin from corn has low molecular weight than chicken egg-derived avidin. This might be due to less complex glycosylation composition in plants. Commercial production of avidin from corn has certain advantages in terms of availability of greater biomass and avoiding the co-purification of animal virus.

Application

Avidin egg white was used in an assay using functionalized xenon as a biosensor to detect biotin-avidin binding. Egg white was used at 80 nmol.
Avidin forms an extremely strong complex with d-biotin (Kd ~ 10-15). This activity has made both avidin and biotin extremely useful labels in immunochemical methods of detection and quantitation.

Unit Definition

One unit will bind 1.0 μg of d-biotin.

Preparation Note

Affinity purified

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ray C Schmidt et al.
Biomaterials, 34(15), 3758-3762 (2013-03-05)
The avidin-biotin system is a highly specific reaction that has been used in a wide range of biomedical applications, including surface modification and cell patterning. We systematically examined a number of avidin derivatives as the basis for a simple and
Tom J A Kokhuis et al.
Ultrasound in medicine & biology, 39(3), 490-506 (2013-01-26)
In this study, we investigated the effect of secondary Bjerknes forces on targeted microbubbles using high-speed optical imaging. We observed that targeted microbubbles attached to an underlying surface and subject to secondary Bjerknes forces deform in the direction of their
Biopharmaceuticals in Plants: Toward the Next Century of Medicine, 121-122 (2009)
Stefan Schillberg et al.
Journal of plant physiology, 258-259, 153359-153359 (2021-01-19)
Recombinant proteins play an important role in many areas of our lives. For example, recombinant enzymes are used in the food and chemical industries and as high-quality proteins for research, diagnostic and therapeutic applications. The production of recombinant proteins is
The avidin-biotin complex (ABC) method and other avidin-biotin binding methods.
G L Bratthauer
Methods in molecular biology (Clifton, N.J.), 115, 203-214 (1999-03-31)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service