Skip to Content
Merck

SA101

Streptavidin

5 mg Streptavidin for ELISA & Western Blotting.

Synonym(s):

Biotin-Binding Protein, Streptavidin Protein

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
NACRES:
NB.22
eCl@ss:
32160405
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


conjugate

unconjugated

Quality Segment

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable, western blot: suitable

shipped in

wet ice

General description

Streptavidin is a tetrameric protein with a globular structure. It is isolated from the bacterium Streptomyces avidinii. Streptavidin is highly thermostable and is resistant to extreme pH, organic solvents, denaturants, detergents, and enzymatic degradation.

Application

Streptavidin has been used to saturate supported lipid bilayers (SLBs) during its preparation.

Biochem/physiol Actions

Streptavidin is widely used as a linker in biotechnology as it forms a very stable bond under well-preserved biological conditions. It exhibits a high binding affinity for biotin. This feature of streptavidin makes it a very efficient and versatile tool for the development of novel sensors. Streptavidin-biotin system is also used in various chemical and biological applications such as detection, labeling, and drug delivery.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library