Skip to Content
Merck
All Photos(1)

Key Documents

A8275

Sigma-Aldrich

Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Goat Anti-Rabbit IgG (whole molecule)–HRP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

peroxidase conjugate

antibody form

IgG fraction of antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct ELISA: 1:10,000

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

IgG, a monomer, is an antibody circulating in blood and can assist in phagocytic destruction of foreign microorganism. It plays an important role in Antibody dependent cell mediated cytotoxicity and is also associated with type II and type III hypersensitivity. Anti-rabbit IgG (whole molecule) peroxidase conjugate antibody can be used in pAb-ELISA. Rabbit anti-IgG antibody coupled with peroxidase (1:2500 dilution in TBS, 0.5% BSA) can be used in western blots. This antibody is also used as detector in indirect ELISA for saxitoxin analysis. Goat anti-rabbit IgG Peroxidase conjugate reacted specifically with all rabbit immunoglobulin.

Immunogen

Purified rabbit IgG.

Application

Anti-rabbit IgG (whole molecule) peroxidase conjugate antibody is used in western blotting, Indirect ELISA, Immunoblotting and Immunodetection.
Co-immunoprecipation and western blot analysis of C33A cell lysates were performed using HRP conjugated goat anti-rabbit IgG as the secondary antibody.
Immunohistochemistry was performed on frozen sections (10um) of mouse intestine, liver, and spleen using HRP-conjugated goat anti-rabbit IgG as the secondary antibody. Prior to incubation with the secondary, sections were treated with a mixture of MeOH/hydrogen peroxide 30% to block endogenouse peroxidases.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4 containing 1% bovine serum albumin with preservative.

Preparation Note

Prepared using the periodate method described by Wilson, M.B., and Nakane, P.K., in Immunofluorescence and Related Staining Techniques, Elsevier/North Holland Biomedical Press, Amsterdam, p215 (1978).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arati Limaye et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 20(7), 959-961 (2006-04-11)
Oral delivery of biopharmaceutical proteins expressed in plant cells should reduce their cost of production, purification, processing, cold storage, transportation, and delivery. However, poor intestinal absorption of intact proteins is a major challenge. To overcome this limitation, we investigate here
Immunoblotting and immunodetection.
Gallagher S, Winston SE, Fuller SA
Current Protocols in Immunology, doi: 10-doi: 10 (2001)
Marta Renato et al.
Plant physiology, 166(2), 920-933 (2014-08-16)
During tomato (Solanum lycopersicum) fruit ripening, chloroplasts differentiate into photosynthetically inactive chromoplasts. It was recently reported that tomato chromoplasts can synthesize ATP through a respiratory process called chromorespiration. Here we show that chromoplast oxygen consumption is stimulated by the electron
Karina Bravo-Tello et al.
PloS one, 12(11), e0187696-e0187696 (2017-11-09)
Soybean meal has been used in many commercial diets for farm fish; despite this component inducing intestinal inflammation. On the other hand, microalgae have increasingly been used as dietary supplements in fish feed. Nevertheless, the vast quantity of microalgae species
L Micheli et al.
Analytical and bioanalytical chemistry, 373(8), 678-684 (2002-08-24)
In this paper the production of antibodies against saxitoxin (STX) is described, as is the optimization and comparison of two competitive ELISA formats (direct and indirect) for the detection of this toxin. Tests were performed in a 96-well microplate using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service