A7510
Adenosine 5′-Triphosphatase from porcine cerebral cortex
lyophilized powder, ≥0.3 units/mg protein, pH 7.8
Synonym(s):
ATP phosphohydrolase, ATPase
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
Porcine cerebral cortex
Quality Level
description
Sodium and Potassium-Activated
form
lyophilized powder
specific activity
≥0.3 units/mg protein, pH 7.8
composition
Protein, 5.0-25.0% biuret
technique(s)
inhibition assay: suitable
storage temp.
−20°C
Gene Information
pig ... ATP1A1(397481) , ATP1B1(396898)
Application
ATPase is used to liberate inorganic phosphorus from ATP. ATPase, from porcine cerebral cortex, has been used in inhibition assays of Na/K-ATPase activity .
Biochem/physiol Actions
This membrane enzyme hydrolyzes ATP to ADP and orthophosphate. The hydrolysis is coupled with the exchange of sodium and potassium ions across the plasma membrane. The enzyme is present in almost all tissues of higher organisms, but it is most abundant in the kidney where it reabsorbs sodium ions from the glomerular filtrate. Na+,K+-ATPase is affected by changes in intracellular calcium. It is also sensitive to ouabain and inhibited by vanadate, and (in vitro) by SPAI-1, a peptide isolated from porcine duodenum. It has been reported to be regulated by phosphorylation by protein kinase A (PKA). Bufalin, and other cardiotonic steroids, may inhibit Na+/K+-ATPase activity and induce the differentiation of HL60, U937, and ML1 cells .
This membrane enzyme hydrolyzes ATP to ADP and orthophosphate. The hydrolysis is coupled with the exchange of sodium and potassium ions across the plasma membrane. The enzyme is present in almost all tissues of higher organisms, but it is most abundant in the kidney where it reabsorbs sodium ions from the glomerular filtrate. Na+,K+-ATPase is affected by changes in intracellular calcium. It is also sensitive to ouabain and inhibited by vanadate, and (in vitro) by SPAI-1, a peptide isolated from porcine duodenum. It has been reported to be regulated by phosphorylation by protein kinase A (PKA).
Unit Definition
One unit will liberate 1.0 μmole of inorganic phosphorus from ATP per min at pH 7.8 at 37 °C in the presence of Na+, K+, and Mg2+.
Physical form
Lyophilized powder containing 90% sucrose, 0.4% EDTA Na4 and 0.06% NaCl
Preparation Note
Sodium iodide extracted fraction of Nakao, et al., Biochem. Biophys. Res. Commun., 19, 755 (1965).
inhibitor
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Nature, 512(7513), 218-222 (2014-07-22)
G-protein-coupled receptors (GPCRs) are critically regulated by β-arrestins, which not only desensitize G-protein signalling but also initiate a G-protein-independent wave of signalling. A recent surge of structural data on a number of GPCRs, including the β2 adrenergic receptor (β2AR)-G-protein complex
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 32(2), 90-100 (2013-12-11)
Clinicopathologic data from a population-based endometrial cancer cohort, unselected for age or family history, were analyzed to determine the optimal scheme for identification of patients with germline mismatch repair (MMR) gene mutations. Endometrial cancers from 702 patients recruited into the
The Journal of cell biology, 205(3), 357-375 (2014-05-07)
The myosin-V family of molecular motors is known to be under sophisticated regulation, but our knowledge of the roles and regulation of myosin-Vs in cytokinesis is limited. Here, we report that the myosin-V Myo51 affects contractile ring assembly and stability
Journal of the National Cancer Institute, 105(4), 274-279 (2013-02-07)
Lynch syndrome is an autosomal dominantly inherited disorder caused by germline mutations in DNA mismatch repair (MMR) genes. Previous studies have shown that MMR gene mutation carriers are at increased risk of colorectal, endometrial, and several other cancers following an
An siRNA screen for NFAT activation identifies septins as coordinators of store-operated Ca2+ entry.
Nature, 499(7457), 238-242 (2013-06-25)
The STIM1-ORAI1 pathway of store-operated Ca(2+) entry is an essential component of cellular Ca(2+) signalling. STIM1 senses depletion of intracellular Ca(2+) stores in response to physiological stimuli, and relocalizes within the endoplasmic reticulum to plasma-membrane-apposed junctions, where it recruits and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service