Skip to Content
Merck
All Photos(4)

Key Documents

A5139

Sigma-Aldrich

Asp-Phe methyl ester

≥98%

Synonym(s):

N-(L-α-Aspartyl)-L-phenylalanine methyl ester, Asp-Phe-OMe, Aspartame

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH2CH(NH2)CONHCH(CH2C6H5)COOCH3
CAS Number:
Molecular Weight:
294.30
Beilstein:
2223850
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.26

Product Name

Asp-Phe methyl ester, ≥98%

Assay

≥98%

form

powder

sweetness

160-200 × sucrose

color

white to off-white

storage temp.

2-8°C

SMILES string

COC(=O)[C@H](Cc1ccccc1)NC(=O)[C@@H](N)CC(O)=O

InChI

1S/C14H18N2O5/c1-21-14(20)11(7-9-5-3-2-4-6-9)16-13(19)10(15)8-12(17)18/h2-6,10-11H,7-8,15H2,1H3,(H,16,19)(H,17,18)/t10-,11-/m0/s1

InChI key

IAOZJIPTCAWIRG-QWRGUYRKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Asp-Phe methyl ester (aspartame, APM, ASP), a dipeptide ester, is made up of phenyl alanine and aspartic acid. Its genotoxic effects have been investigated. Its interaction with certain hydrocolloids has been studied.

Application

Asp-Phe methyl ester is used for the preparation of Ad libitum Standard Chow, with ad libitum drinking water (animals control diet) containing Asp-Phe methyl ester. It is also used for the TFA diet preparation (TFA diet with ad lib drinking water contains Asp-Phe methyl ester.

Biochem/physiol Actions

Asp-Phe methyl ester (Asp-Phe-OMe) is used as a synthetic sweeter, sugar substitute. Asp-Phe methyl ester is being studied for a variety of potential benefits as a nutrition supplement, such as the delay of osteoarthritis and modulation of rheumatoid factor activity. Asp-Phe methyl ester is being studied for its effect on thrombin activity and blood clotting.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interaction of aspartame with selected hydrocolloids: solubility of aspartame.
Wafwoyo W, et al.
Food Hydrocolloids, 13(4), 299-302 (1999)
Kate S Collison et al.
Nutrition & metabolism, 10, 44-44 (2013-06-21)
The human diet has altered markedly during the past four decades, with the introduction of Trans hydrogenated fat, which extended the shelf-life of dietary oils and promoted a dramatic increase in elaidic acid (Trans-18.1) consumption. Food additives such as monosodium
Genotoxicity of aspartame.
Rencuzogullari E, et al.
Drug and Chemical Toxicology, 27(3), 257-268 (2004)
Aspartame: in perspective.
Caseley J and Dixon W.
Nutrition & Food Science, 31(1), 23-27 (2001)
Kate S Collison et al.
Nutrition & metabolism, 9(1), 58-58 (2012-06-16)
Recent evidence suggests that the effects of certain food additives may be synergistic or additive. Aspartame (ASP) and Monosodium Glutamate (MSG) are ubiquitous food additives with a common moiety: both contain acidic amino acids which can act as neurotransmitters, interacting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service