Skip to Content
Merck
All Photos(1)

Key Documents

A5025

Sigma-Aldrich

Ala-Ala-Ala-Ala-Ala

≥99% (TLC)

Synonym(s):

Penta-L-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H27N5O6
CAS Number:
Molecular Weight:
373.40
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

Ala-Ala-Ala-Ala-Ala,

Assay

≥99% (TLC)

Quality Level

form

powder

color

white to off-white

SMILES string

CC(N)C(=O)NC(C)C(=O)NC(C)C(=O)NC(C)C(=O)NC(C)C(O)=O

InChI

1S/C15H27N5O6/c1-6(16)11(21)17-7(2)12(22)18-8(3)13(23)19-9(4)14(24)20-10(5)15(25)26/h6-10H,16H2,1-5H3,(H,17,21)(H,18,22)(H,19,23)(H,20,24)(H,25,26)

InChI key

XXAUOPDVAKGRPR-UHFFFAOYSA-N

Amino Acid Sequence

Ala-Ala-Ala-Ala-Ala

Biochem/physiol Actions

L-Alanine has wide range of applications as food additive and as a component in infusion solutions. It is also used as a precursor for chemical and pharmaceutical products. In a penta-alanine system, resampling helps to increase the efficacy of a “fast-switch” equilibrium sampling protocol.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Resampling improves the efficiency of a ?fast-switch? equilibrium sampling protocol
Biorefineries: Integrated Biochemical Processes for Liquid Biofuels (2009)
Panagiota S Georgoulia et al.
The journal of physical chemistry. B, 115(51), 15221-15227 (2011-11-18)
A computational solution to the protein folding problem is the holy grail of biomolecular simulation and of the corresponding force fields. The complexity of the systems used for folding simulations precludes a direct feedback between the simulations and the force
Edward Lyman et al.
The Journal of chemical physics, 130(8), 081102-081102 (2009-03-05)
We recently applied a multistage reweighting scheme to demonstrate the sampling of equilibrium configurational distributions of peptides from nonequilibrium, simulated annealing trajectories [E. Lyman and D. M. Zuckerman, J. Chem. Phys. 127, 065101 (2007)]. Here we demonstrate that a statistical
Development of growth-arrested bioprocesses with Corynebacterium glutamicum for cellulosic ethanol production from complex sugar mixtures
Biorefineries: Integrated Biochemical Processes for Liquid Biofuels, 121-139 (2014)
Yunfen He et al.
Physical review letters, 101(17), 178103-178103 (2008-11-13)
Terahertz time domain spectroscopy shows that the protein dynamical transition, the rapid increase in protein dynamics occurring at approximately 200 K, needs neither tertiary nor secondary structure. Further, short chain alanine studies find a dynamical transition down to penta-alanine, with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service