Skip to Content
Merck

Skip To

A4487

Aphidicolin

Ready Made Solution, from Nigrospora sphaerica

Synonym(s):

ICI 69653, NSC-234714

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C20H34O4
CAS Number:
Molecular Weight:
338.48
UNSPSC Code:
51102829
NACRES:
NA.85
EC Number:
200-664-3
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Nigrospora sphaerica

Quality Level

vapor pressure

0.55 hPa ( 20 °C)

form

liquid, ready-to-use solution

color

clear colorless

solubility

H2O: miscible (completely)

antibiotic activity spectrum

neoplastics, viruses

mode of action

DNA synthesis | interferes, enzyme | inhibits

storage temp.

−20°C

SMILES string

O[C@]1([C@H]2C[C@@]3([C@@]4([C@H]([C@@]([C@@H](CC4)O)(CO)C)CC[C@H]3C2)C)CC1)CO

InChI

1S/C20H34O4/c1-17(11-21)15-4-3-13-9-14-10-19(13,7-8-20(14,24)12-22)18(15,2)6-5-16(17)23/h13-16,21-24H,3-12H2,1-2H3/t13-,14+,15-,16+,17-,18-,19-,20-/m0/s1

InChI key

NOFOAYPPHIUXJR-APNQCZIXSA-N

General description

Aphidicolin is a tetracyclic diterpene with antiviral and antimitotic properties used for cell cycle synchronization in various cell lines.[1]

Application

Aphidicolin, from Nigrospora sphaerica, was used for cell cycle synchronization of tobacco BY-2 cells. It is used to block the entry of cells into S-phase.[2]

Biochem/physiol Actions

Aphidicolin specifically inhibits DNA polymerase α that is responsible for DNA replication. It also inhibits α-like DNA polymerases of plants and yeasts but does not inhibit synthesis of RNA and proteins. Aphidicolin competes for the dCTP-specific binding site on DNA polymerase α specifically. Aphidicolin has antiviral and antimitotic properties. It blocks the cell cycle at S phase. It has been shown to induce apoptosis in HeLa cells.[3]

Packaging

1 ml

Physical form

Aphidicolin Ready Made Solution supplied as a 1 mg/ml solution in DMSO.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Storage class (TRGS 510): Combustible liquids

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
5.04744SML3001SBR00013
mode of action

DNA synthesis | interferes, enzyme | inhibits

mode of action

-

mode of action

-

mode of action

DNA synthesis | interferes

antibiotic activity spectrum

neoplastics, viruses

antibiotic activity spectrum

-

antibiotic activity spectrum

-

antibiotic activity spectrum

neoplastics

Quality Level

200

Quality Level

100

Quality Level

100

Quality Level

200

form

liquid, ready-to-use solution

form

liquid

form

liquid

form

liquid, ready-to-use solution

solubility

H2O: miscible (completely)

solubility

-

solubility

-

solubility

-

biological source

Nigrospora sphaerica

biological source

-

biological source

-

biological source

Streptomyces sp.


Still not finding the right product?

Explore all of our products under Aphidicolin


Storage Class

10 - Combustible liquids

flash_point_f

188.6 °F - closed cup

flash_point_c

87 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Hui Wang et al.
Cancer research, 77(8), 2148-2160 (2017-02-18)
Overexpression of the EVI1 oncogene is associated typically with aggressive myeloid leukemia, but is also detectable in breast carcinoma where its contributions are unexplored. Analyzing a tissue microarray of 608 breast carcinoma patient specimens, we documented EVI1 overexpression in both
K Marheineke et al.
The Journal of biological chemistry, 276(20), 17092-17100 (2001-03-30)
DNA replication origins are located at random with respect to DNA sequence in Xenopus early embryos and on DNA replicated in Xenopus egg extracts. We have recently shown that origins fire throughout the S phase in Xenopus egg extracts. To
G Pedrali-Noy et al.
Nucleic acids research, 8(2), 377-387 (1980-01-25)
Both the inhibitory effect of aphidicolin on the replicative alpha-polymerase and the reversibility of its action in vivo (Pedrali-Noy & Spadari, 1979, Biochem. Biophys. Res. Commun. 88, 1194-2002) allow the synchronization of cells in culture. Aphidicolin prevents G1 cells from



Global Trade Item Number

SKUGTIN
A4487-1ML04061833374535

Questions

Reviews

No rating value

Active Filters