Skip to Content
Merck
All Photos(1)

Key Documents

A3813

Sigma-Aldrich

Anti-Human Kappa Light Chains (Bound and Free)−Alkaline Phosphatase antibody produced in goat

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Goat Anti-Human Kappa Light Chains (Bound and Free)−AP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

direct ELISA: 1:30,000
dot blot: 1:30,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... IGK@(50802)

General description

Human κ light chains are antibody fragments that regulate immunological responses in cells. Increased expression of κ light chains has been linked to chronic lymphocytic leukemia . The variable fragment of the κ light chain gene has also been associated with vaccine-induced antibody response to the polysaccharide coat in Haemophilus influenzae type b . Anti-Human κ Light Chains (Bound and Free-Alkaline Phosphatase antibody is specific for human κ light chain when tested against human IgA, IgG, IgM, IgGλ, IgAλ, IgMλ, Bence Jones κ and λ myeloma proteins.

Immunogen

Purified Bence Jones κ light chain

Application

Anti-Human κ Light Chains (Bound and Free)-Alkaline Phosphatase antibody is suitable for use in ELISA. The product can also be used for dot blot (1:30,000) and immunohistochemistry (1:50 using formalin-fixed, paraffin-embedded sections).
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)
Mouse-Human chimeric IgG′s specific for Pseudomonas aeruginosa serogroup 06 lipopolysaccharide were identified by ELISA in transfected CHO cell supernatants using Alkaline phosphatase conjugated goat anti-Human Kappa Light chains.

Physical form

Solution in 0.05 M Tris buffer, pH 8.0, containing 1 mM MgCl2, 1% bovine serum albumin, 50% glycerol and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sophie Vazulka et al.
ACS synthetic biology, 11(2), 820-834 (2022-01-19)
Antibody fragments such as Fab's require the formation of disulfide bonds to achieve a proper folding state. During their recombinant, periplasmic expression in Escherichia coli, oxidative folding is mediated by the DsbA/DsbB system in concert with ubiquinone. Thereby, overexpression of
L J Latorre-Rey et al.
Journal of thrombosis and haemostasis : JTH, 15(2), 341-355 (2016-12-09)
Essentials Platelet phenotypes can be modified by lentiviral transduction of hematopoietic stem cells. Megakaryocyte-specific lentiviral vectors were tested in vitro and in vivo for restricted expression. The glycoprotein 6 vector expressed almost exclusively in megakaryocytes. The platelet factor 4 vector
Rogier A van Dijk et al.
Journal of lipid research, 53(12), 2773-2790 (2012-09-13)
The relationships between oxidation-specific epitopes (OSE) and lipoprotein (a) [Lp(a)] and progressive atherosclerosis and plaque rupture have not been determined. Coronary artery sections from sudden death victims and carotid endarterectomy specimens were immunostained for apoB-100, oxidized phospholipids (OxPL), apo(a), malondialdehyde-lysine
Fusion Proteins of Single-Chain Variable Fragments Derived from Phage Display Libraries Are Effective Reagents for Routine Diagnosis of Potato Leafroll Virus Infection in Potato
Toth, R., L., et al.
Virology, 89(11), 1015-1021 (1999)
Thomas Bouquin et al.
Transgenic research, 11(2), 115-122 (2002-06-11)
Transgenic plants represent an alternative to cell culture systems for producing cheap and safe antibodies for diagnostic and therapeutic use. To evaluate the functional properties of a 'plantibody', we generated transgenic Arabidopsis plants expressing full-length human IgG1 against the Rhesus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service