Skip to Content
Merck
All Photos(2)

Key Documents

A2612

Sigma-Aldrich

Asiatic acid

≥98% (HPLC), from Centella asiatica

Synonym(s):

2α,23-Dihydroxyursolic acid, Dammarolic acid, NSC 166063

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H48O5
CAS Number:
Molecular Weight:
488.70
EC Number:
MDL number:
UNSPSC Code:
12352212
PubChem Substance ID:
NACRES:
NA.25

biological source

Centella asiatica

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white

mp

325-330 °C (lit.)

solubility

methanol: 1 mg/mL, clear, colorless

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

C[C@@H]1CC[C@@]2(CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)C[C@@H](O)[C@H](O)[C@@](C)(CO)[C@@H]5CC[C@@]34C)[C@@H]2[C@H]1C)C(O)=O

InChI

1S/C30H48O5/c1-17-9-12-30(25(34)35)14-13-28(5)19(23(30)18(17)2)7-8-22-26(3)15-20(32)24(33)27(4,16-31)21(26)10-11-29(22,28)6/h7,17-18,20-24,31-33H,8-16H2,1-6H3,(H,34,35)/t17-,18+,20-,21-,22-,23+,24+,26+,27+,28-,29-,30+/m1/s1

InChI key

JXSVIVRDWWRQRT-UYDOISQJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Asiatic acid is a naturally occurring pentacyclic triterpenoid, obtained from Centella asiatica.

Application

Asiatic acid has been used in the in vitro study of its cytotoxic potential in glioblastoma cells. It has also been used for the in vivo quantitative analysis of asiatic acid in Centella asiatica L. and in vitro study of enhancement of asiatic acid yield upon using organic elicitors.
Starting material for asiatic acid derivative synthesis for use as:
  • Anticancer agents
  • Glycogen phosphorylase inhibitors
  • Hepatoprotectants

Biochem/physiol Actions

Asiatic acid has been reported to exhibit a pernicious effect on glioblastoma, breast, liver, melanoma, and colon cancers and anti-angiogenic activity. Additionally, it can cross the blood-brain barrier and cause apoptosis in human malignant glioma and hepatoma cells. Asiatic acid also effectively inhibits the Ultraviolet A modulated signal transduction pathways and thereby can be used against UVA-induced photoaging of the skin. It has been noted to show antinociceptive and anti-inflammatory properties plus inhibitory effect on neurotoxicity induced by β-amyloid and glutamate.
Asiatic acid is commonly used in wound healing. Asiatic acid has antioxidant, anti-inflammatory and neuroprotective properties.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantification of asiatic acid from plant parts of Centella asiatica L. and enhancement of its synthesis through organic elicitors in in vitro
Mohammadparast B, et al.,
Horticulture, Environment, and Biotechnology, 55(6), 578-582 (2014)
Byung Chul Park et al.
Cancer letters, 218(1), 81-90 (2005-01-11)
Asiatic acid (AA) is a pentacyclic triterpene found in Centella asiatica. In the present study, the mechanism of anticancer effect of AA on skin cancer was investigated. AA decreased viability and induced apoptosis in human melanoma SK-MEL-2 cells in a
Chandagirikoppal V Kavitha et al.
PloS one, 6(8), e22745-e22745 (2011-08-10)
Malignant gliomas are one of the most devastating and incurable tumors. Sustained excessive angiogenesis by glioma cells is the major reason for their uncontrolled growth and resistance toward conventional therapies resulting in high mortality. Therefore, targeting angiogenesis should be a
Wenjie Guo et al.
PloS one, 7(9), e46018-e46018 (2012-10-03)
Selectively facilitating apoptosis of activated T cells is essential for the clearance of pathogenic injurious cells and subsequent efficient resolution of inflammation. However, few chemicals have been reported to trigger apoptosis of activated T cells for the treatment of hepatitis
Sreenath Nandakumar Nair et al.
Rapid communications in mass spectrometry : RCM, 26(17), 1899-1908 (2012-08-01)
Asiatic acid (AA), a pentacyclic triterpene from Centella asiatica (L.) Urban, has shown numerous therapeutic activities. However, none of the published works to date has used high-performance liquid chromatography/tandem mass spectrometry (HPLC/MS/MS) for determination of AA from biological fluids. Therefore

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service