Skip to Content
Merck
All Photos(1)

Key Documents

A255

Sigma-Aldrich

A-77636 hydrochloride hydrate

≥98% (HPLC), solid

Synonym(s):

(−)-(1R,3S)-3-Adamantyl-1-(aminomethyl)-3,4-dihydro-5,6-dihydroxy-1H-2-benzopyran hydrochloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H27NO3 · HCl · xH2O
CAS Number:
Molecular Weight:
365.89 (anhydrous basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated
protect from light

color

white to off-white

solubility

H2O: >10 mg/mL

storage temp.

−20°C

SMILES string

O.Cl.NC[C@@H]1O[C@@H](Cc2c(O)c(O)ccc12)C34CC5CC(CC(C5)C3)C4

InChI

1S/C20H27NO3.ClH.H2O/c21-10-17-14-1-2-16(22)19(23)15(14)6-18(24-17)20-7-11-3-12(8-20)5-13(4-11)9-20;;/h1-2,11-13,17-18,22-23H,3-10,21H2;1H;1H2/t11?,12?,13?,17-,18-,20?;;/m0../s1

InChI key

KNACNUUSUYTFDY-LIWDBWMRSA-N

Gene Information

human ... DRD1(1812)

Biochem/physiol Actions

Potent, orally active D1 dopamine receptor agonist.

Features and Benefits

This compound is featured on the Dopamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Caution

Hygroscopic, photosensitive, and oxygen-sensitive. Solutions should be freshly prepared.

Legal Information

Manufactured and sold under exclusive license from Abbott Laboratories.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vera Bubenikova-Valesova et al.
The international journal of neuropsychopharmacology, 12(7), 873-883 (2009-01-22)
Cognitive impairment has been found across all subtypes of schizophrenia. The location and function of dopamine-1 receptors (D1Rs) make them attractive targets for the treatment of cognitive impairment in schizophrenia. Here we investigate the systemic effect of a D1R agonist
Allison L Chausmer et al.
Psychopharmacology, 159(2), 145-153 (2002-02-28)
Recent data suggest that dopamine (DA) D1-like receptor full agonists may be potential pharmacotherapeutic agents for treating cocaine abuse. The structurally novel isochroman D1-like agonist, A-77636, has not been well characterized and may prove to be useful as such an
Sanjida S Saklayen et al.
The Journal of pharmacology and experimental therapeutics, 311(1), 342-348 (2004-06-04)
The nigrostriatal dopamine system of the mammalian brain is necessary for normal voluntary motor activity. Dopamine exerts its effects by acting on two primary receptor subtypes: D1-like (D1 and D5) and D2-like (D2, D3, and D4) receptors. Previous research has
S Ofori et al.
General pharmacology, 26(1), 51-57 (1995-01-01)
1. Homogenates of bovine retinas were used to study the capacities of two new dopamine (DA) receptor agonists, e.g. YM 435 and A 77636, to interact with D1 receptors. 2. Adenylate cyclase experiments. YM 435 and A 77636 enhanced cAMP
Amy F T Arnsten et al.
Biological psychiatry, 57(11), 1377-1384 (2005-06-14)
The prefrontal cortex guides behaviors, thoughts, and feelings using representational knowledge, i.e., working memory. These fundamental cognitive abilities subserve the so-called executive functions: the ability to inhibit inappropriate behaviors and thoughts, regulate our attention, monitor our actions, and plan and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service