Skip to Content
Merck
All Photos(2)

Key Documents

A2418

Sigma-Aldrich

Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

IgG fraction of antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse

technique(s)

direct ELISA: 1:15,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:40 using human tonsil tissue
western blot (chemiluminescent): 1:40,000-1:80,000 using deecting β-Actin in total cell extract of HeLa cells (5-10 μg/mL)

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgG (immunoglobulin G) antibody subtype is a predominant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids. Mouse IgGs have four distinct isotypes, namely, IgG1, IgG2a, IgG2b, and IgG3.

Immunogen

purified mouse IgG

Application

Anti-Mouse IgG (whole molecule) Alkaline Phosphatase antibody produced in rabbit has been used in :
  • direct enzyme linked immunosorbent assay (ELISA)
  • immunoblotting
  • immunohistochemistry

Western blot analysis of lysates from the placental cell line 3ASubE-P-3 and U037 cells were performed using alkaline phosphatase conjugated rabbit anti- mouse IgG as the secondary at 1:2000 to recognize the mouse mom Il-8RB. Secondary was developed using bromochloroindoyl phosphate (Sigma) pH. 9.5 for 10-15 minutes.Western blot analysis of coimmunoprecipitates from human cell or rat cell vaginal extracts were performed using alkaline phosphatase conjugated rabbit anti- mouse IgG as the secondary at 1:15000 dilution.

Biochem/physiol Actions

Immunoglobulin G (IgG) is a glycoprotein antibody that regulates immune responses such as phagocytosis and is also involved in the development of autoimmune diseases. IgG antibody participates in complement fixation and opsonization.

Physical form

Solution in 0.05 M Tris, pH 8.0, containing 1% bovine serum albumin, 1 mM MgCl2 and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guangyi Qin et al.
Oncology letters, 15(6), 8756-8760 (2018-05-29)
The present study aimed to investigate the effect of allyl isothiocyanate (AITC) on the viability and apoptosis of the human cervical cancer HeLa cell line in vitro, and to explore the potential underlying mechanisms of this. HeLa cells were treated
Rachel Chikwamba et al.
Transgenic research, 11(5), 479-493 (2002-11-20)
We have produced a functional heat labile enterotoxin (LT-) B subunit of Escherichia coli in maize. LT-B is a multimeric protein that presents an ideal model for an edible vaccine, displaying stability in the gut and inducing mucosal and systemic
Structure and function of human and murine receptors for IgG
Unkeless J C, et al.
Annual Review of Immunology, 6(1), 251-281 (1988)
Dwi Susanti et al.
Frontiers in microbiology, 10, 1256-1256 (2019-07-25)
Phylogenetically deeply rooted methanogens belonging to the genus of Methanocaldococcus living in deep-sea hydrothermal vents derive energy exclusively from hydrogenotrophic methanogenesis, one of the oldest respiratory metabolisms on Earth. These hyperthermophilic, autotrophic archaea synthesize their biomolecules from inorganic substrates and
S El-Amouri et al.
Journal of lipid research, 64(10), 100435-100435 (2023-08-29)
Obesity-associated diabetes is linked to the accumulation of ceramide in various organs, including the liver. The exact mechanisms by which ceramide contributes to diabetic pathology are unclear, but one proposed scenario is that ceramide accumulation may inhibit insulin signaling pathways.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service