Skip to Content
Merck
All Photos(1)

Key Documents

PHR1654

Supelco

Amikacin

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Amikacin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H43N5O13
CAS Number:
Molecular Weight:
585.60
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur.
traceable to USP 1019508

API family

amikacin

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N([C@H]1[C@@H]([C@H]([C@@H]([C@H](C1)N)O[C@H]3O[C@@H]([C@H]([C@@H]([C@H]3O)O)O)CN)O)O[C@H]2O[C@@H]([C@H]([C@@H]([C@H]2O)N)O)CO)C(=O)[C@@H](O)CCN

InChI

1S/C22H43N5O13/c23-2-1-8(29)20(36)27-7-3-6(25)18(39-22-16(34)15(33)13(31)9(4-24)37-22)17(35)19(7)40-21-14(32)11(26)12(30)10(5-28)38-21/h6-19,21-22,28-35H,1-5,23-26H2,(H,27,36)/t6-,7+,8-,9+,10+,11-,12+,13+,14+,15-,16+,17-,18+,19-,21+,22+/m0/s1

InChI key

LKCWBDHBTVXHDL-RMDFUYIESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Amikacin is an aminoglycoside antibiotic. It is a semisynthetic analog of kanamycin and is effective against most gram-negative bacteria. It is used for the treatment of skin infections.
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

Amikacin may be used as a pharmaceutical secondary reference standard for the determination of the analyte in plasma and milk samples by various chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB9944 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of amikacin in human plasma by high-performance capillary electrophoresis with fluorescence detection
Oguri S and Miki Y
Journal of Chromatography. B, Biomedical Sciences and Applications, 686(2), 205-210 (1996)
Amikacin serum concentrations: prediction of levels and dosage guidelines
Sarubbi FA, et al.
Annals of Internal Medicine, 89, 612-618 (1978)
Amikacin serum concentrations: prediction of levels and dosage guidelines
Sarubbi FA, et al.
Annals of Internal Medicine, 89(5-1), 612-618 (1978)
Assay of amikacin in the skin by high-performance liquid chromatography
Nicoli S and Santi P
Journal of Pharmaceutical and Biomedical Analysis, 41(3), 994-997 (2006)
Determination of kanamycin A, amikacin and tobramycin residues in milk by capillary zone electrophoresis with post-column derivatization and laser-induced fluorescence detection
Yu CZ, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 877(3), 333-338 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service