Skip to Content
Merck
All Photos(1)

Key Documents

A0637

Sigma-Aldrich

DL-2-Aminoadipic acid

≥99% (TLC), powder, gliotoxic

Synonym(s):

DL-α-Aminoadipic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2C(CH2)3CH(NH2)CO2H
CAS Number:
Molecular Weight:
161.16
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

Product Name

DL-2-Aminoadipic acid, ≥99%

Quality Level

Assay

≥99%

form

powder

mp

196-198 °C (lit.)

storage temp.

2-8°C

SMILES string

NC(CCCC(O)=O)C(O)=O

InChI

1S/C6H11NO4/c7-4(6(10)11)2-1-3-5(8)9/h4H,1-3,7H2,(H,8,9)(H,10,11)

InChI key

OYIFNHCXNCRBQI-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

DL-2-Aminoadipic acid (AAA) has been used as an astrotoxin to kill astrocytes that prevent efficient integration of transplanted cells into the retina. It has also been used as a gliotoxin to study the effect of astrocytic swelling on the tortuosity.[1]

Biochem/physiol Actions

DL-2-Aminoadipic acid (AAA) is a six-carbon homolog of glutamate[2] and a gliotoxic compound.[3] It is generally used to considerably reduce the number of astroglia in cerebellar cultures that acts as a model to study the mechanisms of a-aminoadipic acid induced glial toxicity.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gliotoxic effects of $\alpha$-aminoadipic acid on monolayer cultures of dissociated postnatal mouse cerebellum
Huck S, et al.
Neuroscience, 12(3), 783-791 (1984)
Gliotoxin-induced swelling of astrocytes hinders diffusion in brain extracellular space via formation of dead-space microdomains
Sherpa AD, et al.
Glia, 62(7), 1053-1065 (2014)
H Q Wu et al.
European journal of pharmacology, 281(1), 55-61 (1995-07-25)
L-alpha-Aminoadipic acid is a lysine metabolite with neuroexcitatory properties, and has previously been shown to inhibit the production of the broad spectrum excitatory amino acid receptor antagonist kynurenic acid in brain tissue slices. The effects of L-alpha-aminoadipic acid on the
Huadong Ni et al.
Journal of neuroinflammation, 16(1), 1-1 (2019-01-05)
Despite accumulating evidence on the role of glial cells and their associated chemicals in mechanisms of pain, few studies have addressed the potential role of chemokines in the descending facilitation of chronic pain. We aimed to study the hypothesis that
The toxic effect of sodium glutamate and DL-$\alpha$-aminoadipic acid on rat retina: Changes in high affinity uptake of putative transmitters
Karlsen RL, et al.
Journal of Neurochemistry, 31(4), 1055-1061 (1978)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service