Skip to Content
Merck
All Photos(1)

Key Documents

A0487

Sigma-Aldrich

Argatroban monohydrate

≥98% (HPLC)

Synonym(s):

(2R,4R)-1-[(2S)-5-[(Aminoiminomethyl)amino]-1-oxo-2-[[(1,2,3,4-tetrahydro-3-methyl-8-quinolinyl)sulfonyl]amino]pentyl]-4-methyl-2-piperidinecarboxylic Acid, Argipidine, MQPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H36N6O5S · H2O
CAS Number:
Molecular Weight:
526.65
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to off-white

solubility

DMSO: ≥20 mg/mL

originator

Baxter

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(NC(CCCNC(=N)N)C(=O)N3[C@H](C[C@@H](CC3)C)C(=O)O)c1c2c(ccc1)CC(CN2)C.O

InChI

1S/C23H36N6O5S.H2O/c1-14-8-10-29(18(12-14)22(31)32)21(30)17(6-4-9-26-23(24)25)28-35(33,34)19-7-3-5-16-11-15(2)13-27-20(16)19;/h3,5,7,14-15,17-18,27-28H,4,6,8-13H2,1-2H3,(H,31,32)(H4,24,25,26);1H2/t14-,15?,17?,18-;/m1./s1

InChI key

AIEZTKLTLCMZIA-LINCPPCXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Argatroban anticoagulant is synthesized from L-arginine and photodegrades under light exposure.

Application

Argatroban monohydrate has been used as a thrombin-selective inhibitor in the thrombin assay of platelet poor plasma samples. It may be used as a standard in gradient reversed-phase liquid chromatography coupled with high-resolution multistage mass spectrometry (LC/HR-MSn) for photoproducts fragmentation analysis and as a thrombin inhibitor competitive assay with fullerene-based C60 nanoparticle (NPs).

Biochem/physiol Actions

Argatroban has a high affinity and binds to arginine in thrombin active site resulting in steric hindrance for substrate binding. Argatroban is effective in preventing thrombus formation and favors thrombolysis than unfractionated heparin. It is recommended for acute ischemic stroke treatment.
Argatroban is a potent, selective, direct inhibitor of thrombin and a clinical anticoagulant.
Argatroban is a potent, seletive, univalent direct inhibitor of thrombin. It directly inhibits thrombin by binding only to its active site (thus univalent) as compared to Bivalent DTIs (hirudin and analogs) which bind both to the active site and exosite 1.

Features and Benefits

This compound was developed by Baxter. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arnat Balabiyev et al.
Biomaterials, 277, 121087-121087 (2021-09-04)
Implantation of biomaterials and medical devices in the body triggers the foreign body reaction (FBR) which is characterized by macrophage fusion at the implant surface leading to the formation of foreign body giant cells and the development of the fibrous
Photodegradation of aqueous argatroban investigated by LC/MSn: Photoproducts, transformation processes and potential implications
Secretan PH, et al.
Journal of Pharmaceutical and Biomedical Analysis, 131(29387), 223-232 (2016)
Solution-Mediated Phase Transformation of Argatroban: Ternary Phase Diagram, Rate-Determining Step, and Transformation Kinetics
Wang Y, et al.
Industrial & Engineering Chemistry Research, 56(15), 4539-4548 (2017)
Jeremy B Chang et al.
Scientific reports, 6, 29387-29387 (2016-07-22)
A central challenge in designing and administering effective anticoagulants is achieving the proper therapeutic window and dosage for each patient. The Hill coefficient, nH, which measures the steepness of a dose-response relationship, may be a useful gauge of this therapeutic
Effects of argatroban and heparin on thrombus formation and tissue plasminogen activator-induced thrombolysis in a microvascular thrombosis model
Yamada K, et al.
Thrombosis Research, 109(1), 55-64 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service