Skip to Content
Merck
All Photos(3)

Key Documents

A0418

Sigma-Aldrich

Anti-Rabbit IgG (whole molecule)–Alkaline Phosphatase antibody produced in goat

IgG fraction of antiserum

Synonym(s):

Goat Anti-Rabbit IgG (whole molecule)–AP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

IgG fraction of antiserum

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

rabbit

technique(s)

direct ELISA: 1:20,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:40
western blot (chemiluminescent): 1:100,000

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgG crosses the placental barrier, is a complement activator and binds to the Fc-receptors on phagocytic cells. The level of IgG may vary with the status of disease or infection. Alkaline Phosphatase is an enzyme that catalyzes the conversion of chromogenic substrates such as p-nitrophenylphosphate (PNPP); chemiluminescent substrates such as CDP-Star® and fluorogenic substrates such as 4-methylumbelliferyl phosphate (4-MUP) into detectable chromophores, light-emitters or fluorescers, respectively.
Rabbit IgG is a plasma B cell derived antibody isotype defined by its heavy chain. IgG is the most abundant antibody isotype found in rabbit serum. Immunoglobulins (Igs) belongs to the immunoglobulin super-family. Each immunoglobin have two heavy (H) and two light (L) chains, held together by disulphide linkages. Each light chain comprises one variable N-terminal region and a constant C-terminal region. Heavy chain has one variable N-terminal region and three or four constant (CH1-CH4) C-terminal region. The four classes of IgG can be IgG1, IgG2, IgG3, and IgG4.

Specificity

Goat polyclonal anti-rabbit IgG (whole molecule)-alkaline phosphatase antibody reacts with rabbit IgG in vitro, in rabbit serum and biological fluids.

Immunogen

pooled normal rabbit serum

Application

Anti-Rabbit IgG (whole molecule)−Alkaline Phosphatase antibody produced in goat has been used in western blotting and enzyme-linked immunosorbent assay (ELISA).
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)
Citrullination of antithrombin by PADI4 was analyzed by ELISA using alkaline phosphatase conjugated goat anti-rabbit IgG as the secondary diluted at 1:5000 in 0.05M carbonate/bicarbonate buffer (Ph 9.6).

Biochem/physiol Actions

IgG1 is most abundant and its deficiency results in hypogammaglobulinemia. IgG2 deficiency increases susceptibility to bacterial infections. IgG3 mediates effector functions and IgG4 is associated with asymptomatic infection. IgG antibody have enormous therapeutic potential and the Fc region contributes for the development of therapeutic antibody.

Physical form

Solution in 0.05 M Tris buffer, pH 8.0, containing 1% bovine serum albumin, 1 mM MgCl2 and 15 mM sodium azide

Legal Information

CDP-Star is a registered trademark of Tropix, Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CIM monolith chromatography-enhanced ELISA detection of proteins in artists' paints: Ovalbumin as a case study
Spec T, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 127, 102-112 (2016)
Sissi Miguel et al.
Communications biology, 3(1), 673-673 (2020-11-15)
The synthesis of 3,5-dicaffeoylquinic acid (3,5-DiCQA) has attracted the interest of many researchers for more than 30 years. Recently, enzymes belonging to the BAHD acyltransferase family were shown to mediate its synthesis, albeit with notably low efficiency. In this study
Carmel M McVicar et al.
Fertility and sterility, 81 Suppl 1, 767-774 (2004-03-17)
To determine the incidence of Fas positivity and DNA double-strand breaks (DSB) as indicators of early- and late-stage apoptosis in ejaculated sperm. Fas positivity was assessed by flow cytometry and DSB by the neutral Comet assay. Andrology Laboratory, Royal Maternity
Assessing carnivorous plants for the production of re-combinant proteins
Miguel S, et al.
Frontiers in Plant Science, 10, 793-793 (2019)
Cellular casein kinase 2 and protein phosphatase 2A modulate replication site assembly of bluetongue virus
Mohl BP and Roy P
Test, 291(28), 14566-14574 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service