Skip to Content
Merck
All Photos(1)

Key Documents

A0231

Sigma-Aldrich

AS 604850

≥98% (HPLC), solid

Synonym(s):

5-(2,2-Difluoro-benzo[1,3]dioxol-5-ylmethylene)-thiazolidine-2,4-dione, PI3Kγ inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H5F2NO4S
CAS Number:
Molecular Weight:
285.22
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

off-white

solubility

DMSO: >10 mg/mL
H2O: <2 mg/mL

shipped in

wet ice

storage temp.

2-8°C

SMILES string

FC1(F)Oc2ccc(cc2O1)\C=C3/SC(=O)NC3=O

InChI

1S/C11H5F2NO4S/c12-11(13)17-6-2-1-5(3-7(6)18-11)4-8-9(15)14-10(16)19-8/h1-4H,(H,14,15,16)/b8-4-

InChI key

SRLVNYDXMUGOFI-YWEYNIOJSA-N

Biochem/physiol Actions

AS 604850 is a selective PI3Kγ inhibitor

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Phosphoinositide Kinases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniela Kovacs et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(5), 6302-6321 (2020-03-12)
Bovine colostrum, the first milk secreted by the mammary glands of cows shortly after they have given birth, provides a natural source of bioactive substances helpful to promote tissue development and repair, and to maintain a healthy immune system. Owing
Montserrat Camps et al.
Nature medicine, 11(9), 936-943 (2005-08-30)
Phosphoinositide 3-kinases (PI3K) have long been considered promising drug targets for the treatment of inflammatory and autoimmune disorders as well as cancer and cardiovascular diseases. But the lack of specificity, isoform selectivity and poor biopharmaceutical profile of PI3K inhibitors have
Chatvadee Kornsuthisopon et al.
Scientific reports, 14(1), 6777-6777 (2024-03-22)
Extracellular matrix (ECM) is an intricate structure providing the microenvironment niche that influences stem cell differentiation. This study aimed to investigate the efficacy of decellularized ECM derived from human dental pulp stem cells (dECM_DPSCs) and gingival-derived mesenchymal stem cells (dECM_GSCs)
Hon Yan K Yip et al.
Molecular cell, 80(2), 279-295 (2020-10-17)
The PTEN tumor suppressor controls cell death and survival by regulating functions of various molecular targets. While the role of PTEN lipid-phosphatase activity on PtdIns(3,4,5)P3 and inhibition of PI3K pathway is well characterized, the biological relevance of PTEN protein-phosphatase activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service